Skip to Content
Merck
All Photos(1)

Key Documents

I4758

Sigma-Aldrich

Iminodiacetic acid Agarose

saline suspension

Synonym(s):

IDA Resin

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$239.00
50 ML
$1,670.00

$239.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
5 ML
$239.00
50 ML
$1,670.00

About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.56

$239.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


form

saline suspension

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

epoxy

matrix attachment

amino

matrix spacer

12 atoms

capacity

≥15 μmol/mL binding capacity (CuSO4)

storage temp.

2-8°C

SMILES string

[X]N(CC(=O)O)CC(=O)O

Looking for similar products? Visit Product Comparison Guide

Application

Iminodiacetic acid-agarose is used in affinity chromatography, protein chromatography and as a chelating resin.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Zatloukalová et al.
Journal of separation science, 29(8), 1082-1087 (2006-07-13)
A method for the immobilization of antibodies to inert matrix represents an important factor that affects results of immunoaffinity chromatography. Binding antibodies to immobilized metal ions is an example of oriented immobilization that avoids a random coupling of a protein.
S Ghosh et al.
Gene, 176(1-2), 249-255 (1996-10-17)
Vectors have been constructed for the general purpose of expressing foreign proteins in E. coli. These vectors allow the production in high yield of either native proteins or of fusion proteins which contain, at their amino terminus, the peptide Met
S S Lam et al.
Biochemical and biophysical research communications, 253(1), 135-142 (1999-01-06)
Two ribosome inactivating proteins designated alpha- and beta-pisavins were isolated from seeds of the garden pea Pisum sativum var. arvense Poir with a procedure involving affinity chromatography on Affi-gel Blue gel, immobilized metal ion affinity chromatography on Iminodiacetic acid-agarose, cation

Questions

  1. What is the maximum flow rate for iminodiacetic acid agarose (#I4758)?

    1 answer
    1. Product I4758 is 4% beaded agarose with a particle size of 45-165μm. For these products, a maximum operating pressure of 0.08 MPa and a maximum flow rate of 1.0mL/min are recommended.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service