Skip to Content
Merck
All Photos(1)

Key Documents

I3631

Sigma-Aldrich

Isopentyl nitrite

Synonym(s):

Isoamyl nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2ONO
CAS Number:
Molecular Weight:
117.15
Beilstein:
969510
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

refractive index

n20/D 1.386 (lit.)

bp

99 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCON=O

InChI

1S/C5H11NO2/c1-5(2)3-4-8-6-7/h5H,3-4H2,1-2H3

InChI key

OWFXIOWLTKNBAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 150495-Sigma-Aldrich | Isopentyl nitrite 96%

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hossein Mehrabi
Ultrasonics sonochemistry, 15(4), 279-282 (2007-10-26)
The reaction of 6-acylmethylphenanthridines with isoamyl nitrite results alpha-oximino-6-acylmethylphenanthridines in 73-95% yields in DMF under ultrasound irradiation. Compared with conventional methods, the main advantages of the present procedure are milder conditions, shorter reaction time and higher yields.
Analysis of alkyl nitrites by capillary gas chromatography-mass spectrometry.
T S Bal et al.
Journal - Forensic Science Society, 28(3), 185-190 (1988-05-01)
R M Uppu et al.
Analytical biochemistry, 236(2), 242-249 (1996-05-01)
A new method for the preparation of high concentrations of peroxynitrite (up to 1 M) is described. The synthesis uses a two-phase system and involves a displacement reaction by the hydroperoxide anion (in the aqueous phase) on isoamyl nitrite (in
K Reszka et al.
Free radical research communications, 10(1-2), 47-56 (1990-01-01)
Diazotization of primary aromatic amines with isoamyl nitrite in benzene at room temperature was studied employing EPR and spin trapping techniques. Nitrosodurene (ND), 2-methyl-2-nitrosopropane (MNP), and 5,5-dimethyl-pyrroline N-oxide (DMPO) were used as spin trapping agents. Aryl radicals were detected employing
Rao M Uppu
Analytical biochemistry, 354(2), 165-168 (2006-06-06)
A method for the synthesis of peroxynitrite is described. It involves nitrosation of H2O2 at pH> or = 12.5 by isoamyl or butyl nitrite in mixed solvents of isopropyl alcohol (IPA) and water at 25+/-1 degrees C. Maximum yields of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service