Skip to Content
Merck
All Photos(1)

Key Documents

I2911

Sigma-Aldrich

IMDM

StableCell, with stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Iscove’s Modified Dulbecco’s Medium, IMDM

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$68.90

$68.90


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$68.90

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

$68.90


Estimated to ship onMay 28, 2025


Request a Bulk Order

Product Name

StableCell IMDM, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

NaHCO3: yes
glucose: yes
stable glutamine: yes
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This IMDM differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.

Application

Guilbert and Iscove demonstrated that precursor cells of erythrocytes and macrophages could be cultured in a totally defined serum free medium supplemented with albumin, transferrin, lecithin, and selenium. This medium is a modification of Dulbecco′s Modified Eagle′s Medium (DME) containing selenium, additional amino acids and vitamins, sodium pyruvate, HEPES buffer, and potassium nitrate instead of ferric nitrate. Further studies demonstrated that Iscove′s Medium would support murine B lymphocytes, hemopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleni Gelali et al.
Nature communications, 10(1), 1636-1636 (2019-04-11)
DNA fluorescence in situ hybridization (DNA FISH) is a powerful method to study chromosomal organization in single cells. At present, there is a lack of free resources of DNA FISH probes and probe design tools which can be readily applied.
Gustav Nilsonne et al.
Behavioral and brain functions : BBF, 7, 47-47 (2011-11-22)
Recent advances in cell biology and gene regulation suggest mechanisms whereby associative learning could be performed by single cells. Therefore, we explored a model of classical conditioning in human macrophages in vitro. In macrophage cultures, bacterial lipopolysaccharide (LPS; unconditioned stimulus)
Christine Bourgeois et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(25), 8691-8696 (2008-06-20)
A model of chemical thymectomy by inducible Rag ablation was used to study peripheral T cell homeostasis. Induction of Rag ablation was efficient and complete, leading to cessation of thymic T cell production within 3-4 weeks. The decay of peripheral
Florian A Salomons et al.
Molecular and cellular biology, 29(7), 1774-1785 (2009-01-23)
Conditions causing an increase in misfolded or aberrant proteins can impair the activity of the ubiquitin/proteasome system (UPS). This observation is of particular interest, given the fact that proteotoxic stress is closely associated with a large variety of disorders. Although

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service