Skip to Content
Merck
All Photos(1)

Key Documents

I2411

Sigma-Aldrich

Isocitrate Dehydrogenase 1 (NADP+) human

recombinant, expressed in Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

IDH1, Isocitrate Dehydrogenase Cytoplasmic

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$900.00

$900.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
10 μG
$900.00

About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

$900.00


Estimated to ship onMay 30, 2025


recombinant

expressed in Sf9 cells

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

47.6 kDa

concentration

≥0.30 mg/mL protein (Bradford)

NCBI accession no.

relevant disease(s)

cancer (lung)

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Human Isocitrate Dehydrogenase (IDH1), also known as oxalosuccinate decarboxylase, full length [amino acids 1-414 (end)] with C-terminal FLAG-tag, expressed in Sf9 cells via a baculovirus expression system.

Application

Human Isocitrate Dehydrogenase (IDH1) is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. It may be useful as a biomarker for diagnosis and prognosis prediction of non-small cell lung cancer [1].

Biochem/physiol Actions

Human Isocitrate Dehydrogenase (IDH1), also known as oxalosuccinate decarboxylase, is a full length amino acid with a C-terminal FLAG-tag and is expressed in Sf9 cells via a baculovirus expression system. IDH1 expression has been correlated with poor survival of non-small cell lung cancer. IDH1 promotes tumor growth [1].

Physical form

Solution in 45 mM Tris-HCl, 124 mM NaCl, 2.4 mM KCl, 90 μg/ml FLAG peptide, 3 mM DTT, and 10% glycerol at pH 8.0.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengwei Tan et al.
Molecular & cellular proteomics : MCP, 11(2), M111-M111 (2011-11-09)
Lung cancer is the leading cause of cancer-related death in the world. To explore tumor biomarkers for clinical application, two-dimensional fluorescence difference gel electrophoresis and subsequent MALDI-TOF/TOF mass spectrometry were performed to identify proteins differentially expressed in 12 pairs of
Peter Paschka et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(22), 3636-3643 (2010-06-23)
To analyze the frequency and prognostic impact of isocitrate dehydrogenase 1 (IDH1) and isocitrate dehydrogenase 2 (IDH2) mutations in acute myeloid leukemia (AML). We studied 805 adults (age range, 16 to 60 years) with AML enrolled on German-Austrian AML Study
Lenny Dang et al.
Trends in molecular medicine, 16(9), 387-397 (2010-08-10)
The systematic sequencing of glioblastoma multiforme (GBM) genomes has identified the recurrent mutation of IDH1, a gene encoding NADP(+)-dependent isocitrate dehydrogenase 1 (IDH1) that catalyzes the oxidative decarboxylation of isocitrate yielding alpha-ketoglutarate (alpha-KG). Subsequent studies have confirmed recurrent IDH1 and
Yuejun Fu et al.
Biochemical and biophysical research communications, 397(2), 127-130 (2010-06-01)
Heterozygous mutations in either the R132 residue of isocitrate dehydrogenase I (IDH1) or the R172 residue of IDH2 in human gliomas were recently highlighted. Heterozygous mutations in the IDH1 occur in the majority of grade II and grade III gliomas
Matthew J Schniederjan et al.
The American journal of surgical pathology, 37(5), 763-771 (2013-04-17)
Leptomeningeal dissemination in children is typical of high-grade, and occasionally low-grade, neoplasms. Rare cases of widely disseminated oligodendroglia-like leptomeningeal tumors, sometimes with associated spinal cord lesions, have been described that respond to treatment and follow an indolent course. Whether these

Protocols

We describe here a rapid and sensitive method to separate and measure D-2-OHG and L-2-OHG enantiomers using high-resolution mass spectrometry (HRMS) detection.

Chromatograms

application for HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service