Skip to Content
Merck
All Photos(2)

Documents

I1659

Sigma-Aldrich

Anti-IFI-16 antibody, Mouse monoclonal

~1 mg/mL, clone IFI-230, purified from hybridoma cell culture

Synonym(s):

Anti-Interferon-inducible 16

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

IFI-230, monoclonal

form

buffered aqueous solution

mol wt

antigen 85-95 kDa (cluster of three isotypes A, B, and C)

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using whole extract of the Jurkat cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFI16(3428)

General description

The gene IFI-16 (interferon, γ-inducible protein 16) encodes a member of the interferon-inducible nuclear proteins family containing one or two copies of a conserved 200 amino acid repeat domain called HIN-200 domain. It has three different isoforms, IFI-16A, -16B, -16C. The IFI-16A isoform is the longest with 785 amino acids. IFI-16B is the most broadly expressed isoform with 729 amino acids and IFI-16C has 56 amino acids lesser. The three isoforms have serine and threonine residues which can be phosphorylated. The N-terminus contains an imperfect leucine zipper domain, where the isoforms homo- and heterodimerize. IFI-16 contains an N-terminal domain called DAPIN/PYRIN/CARD that may be involved in protein–protein interactions. IFI-16 is expressed in some haemopoietic lineages , such as CD34+ progenitor cells, mature lymphocytes and monocytes. The gene is mapped to human chromosome 1q22.

Immunogen

synthetic peptide corresponding to amino acids 768-783 at the C-terminus of human IFI-16.

Biochem/physiol Actions

The gene IFI-16 (interferon, γ-inducible protein 16) encodes an interferon-inducible transcriptional modulator from the p200-protein family. It may play a role in negative regulation of transcription. Overexpression of this gene in older populations of human fibroblasts leads to cellular senescence. It is also associated with senescence-associated irreversible cell growth arrest in normal human prostate epithelial cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brendan Antiochos et al.
JCI insight, 3(18) (2018-09-21)
IFN-inducible protein 16 (IFI16) is an innate immune sensor that forms filamentous oligomers when activated by double-stranded DNA (dsDNA). Anti-IFI16 autoantibodies occur in patients with Sjögren's syndrome (SS) and associate with severe phenotypic features. We undertook this study to determine
Hong Xin et al.
Oncogene, 23(37), 6209-6217 (2004-06-23)
Defects in interferon (IFN) signaling that result in loss of expression of IFN-inducible proteins are associated with cellular immortalization, an important early event in the development of human cancer. Here we report that loss of IFN-inducible IFI 16 expression in
Wu Wei et al.
Histochemistry and cell biology, 119(1), 45-54 (2003-01-28)
IFI 16 is a member of the HIN-200 protein family named for their haemopoietic expression, interferon-inducibility and nuclear localisation. These proteins have been characterised as transcriptional regulators that modulate the cell cycle. IFI 16 is expressed in some haemopoietic lineages
R W Johnstone et al.
Biochemistry, 37(34), 11924-11931 (1998-08-26)
We recently demonstrated that IFI 16, a human member of a family of interferon-inducible nuclear proteins, can function as a potent repressor of transcription. All members of this family are found in the nucleus and contain 1 or 2 copies
R W Johnstone et al.
The Journal of biological chemistry, 273(27), 17172-17177 (1998-06-27)
IFI 16 is a member of a family of interferon-inducible proteins, including the human MNDA (myeloid nuclear differentiation antigen), the recently identified AIM-2 (absent in melanoma), and the homologous murine molecules, p202, p204, and D3. IFI 16 contains a domain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service