Skip to Content
Merck
All Photos(1)

Key Documents

I1392

Sigma-Aldrich

HA-100

Synonym(s):

1-(5-Isoquinolinesulfonyl)piperazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N3O2S
CAS Number:
Molecular Weight:
277.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.O=S(=O)(N1CCNCC1)c2cccc3cnccc23

Gene Information

Biochem/physiol Actions

Inhibitor of PKA, PKC and myosin light chain kinase.

Quality

HCl content may vary.

Analysis Note

Actual content given on the label

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J T O'Flaherty et al.
Biochemical and biophysical research communications, 163(3), 1456-1460 (1989-09-29)
Three protein kinase C blockers (staurosporin, Cl, and sphinganine) acted temperature- and time-dependently on human neutrophils to lower the affinity and number of high affinity plasmalemma receptors available to leukotriene B4. The drugs did not alter the ligand's binding to
D A Bass et al.
The Journal of biological chemistry, 262(14), 6643-6649 (1987-05-15)
Preincubation of neutrophils with certain agonists may "prime" the cells to cause increased responses to a second stimulus ("primed stimulation"). We used two approaches to examine the role of protein kinase C (Ca2+/phospholipid-dependent enzyme) in priming and stimulation by 1-oleoyl-2-acetylglycerol
L Harvath et al.
Journal of immunology (Baltimore, Md. : 1950), 139(9), 3055-3061 (1987-11-01)
The protein kinase inhibitor, 1-(5-isoquinolinesulfonyl) piperazine (C-I), inhibits superoxide release from human neutrophils (PMN) stimulated with phorbol myristate acetate or synthetic diacylglycerol, without inhibiting superoxide release from PMN stimulated with the chemoattractants C5a or N-formyl-methionyl-leucyl-phenylalanine (f-Met-Leu-Phe). In this study, we
M Awazu et al.
Journal of cardiovascular pharmacology, 17 Suppl 7, S500-S502 (1991-01-01)
The vasoconstrictor response is defective in diabetes mellitus (DM). Activation of protein kinase C (PKC) is also known to prevail in diabetes mellitus, and it is thought to be secondary to abnormal diacylglycerol metabolism. To ascertain whether this PKC activation
Regulation of neutrophil chemotaxis: correlations with actin polymerization.
L Harvath
Cancer investigation, 8(6), 651-654 (1990-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service