Skip to Content
Merck
All Photos(1)

Key Documents

I0661

Sigma-Aldrich

(−)-Indolactam V

≥96% (HPLC)

Synonym(s):

(2S,5S)-1,2,4,5,6,8-Hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3H-pyrrolo[4,3,2-gh]-1,4-benzodiazonin-3-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$444.00

$444.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$444.00

About This Item

Empirical Formula (Hill Notation):
C17H23N3O2
CAS Number:
Molecular Weight:
301.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$444.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

Assay

≥96% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)[C@@H]1N(C)c2cccc3[nH]cc(C[C@@H](CO)NC1=O)c23

InChI

1S/C17H23N3O2/c1-10(2)16-17(22)19-12(9-21)7-11-8-18-13-5-4-6-14(15(11)13)20(16)3/h4-6,8,10,12,16,18,21H,7,9H2,1-3H3,(H,19,22)/t12-,16-/m0/s1

InChI key

LUZOFMGZMUZSSK-LRDDRELGSA-N

Biochem/physiol Actions

(-)-Indolactam V is a PKC activator shown to effect differentiation in embryonic stem cells leading to development of pancreatic precursors.
(-)-Indolactam V is a PKC activator shown to effect differentiation in embryonic stem cells leading to development of pancreatic precursors. It is active in the mouse model.
(−)-Indolactam V is naturally present in Streptoverticillium blastmyceticum.[1] It serves as a precursor to the teleocidin class which are alkaloids.[2] It is a the pharmacophore of lyngbyatoxin[1] and teleocidins.[2]

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengren Xu et al.
Organic & biomolecular chemistry, 9(7), 2512-2517 (2011-02-23)
The total synthesis of protein kinase C activator (-)-indolactam V (IL-V) has been successfully completed with two separate approaches: From known 4-nitrotryptophan derivative 3 in 8 steps (49% overall yield) and from L-glutamic acid in 12 steps (18% overall yield)
Kazuhiro Irie et al.
Chemical record (New York, N.Y.), 5(4), 185-195 (2005-07-26)
Tumor promoters such as phorbol esters bind strongly to protein kinase C (PKC) isozymes to induce their activation. Since each PKC isozyme is involved in diverse biological events in addition to tumor promotion, the isozymes serve as promising therapeutic targets.
Synthesis of (-)-Epi-Indolactam V by an Intramolecular Buchwald-Hartwig C-N Coupling Cyclization Reaction
Mari M, et al.
The Journal of Organic Chemistry, 78(15), 7727-7734 (2013)
Daniel J Edwards et al.
Journal of the American Chemical Society, 126(37), 11432-11433 (2004-09-16)
The lyngbyatoxins are potent skin irritants produced by Lyngbya majuscula and cause a condition known as "Swimmer's Itch" off Honolulu, HI. Reported is the molecular cloning of the lyngbyatoxin (ltx) biosynthetic gene cluster from L. majuscula using a strategy based
Kazuhiro Irie et al.
Bioorganic & medicinal chemistry, 12(17), 4575-4583 (2004-09-11)
Recent investigations discovered nonkinase-type phorbol ester receptors, RasGRPs, chimaerins, and Unc13s. Phorbol ester binding occurs at the cysteine-rich sequences of about 50 residues in the C1 domains of these receptors. Fifty-one-residue RasGRP C1 peptides except for RasGRP2 showed significant phorbol

Articles

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service