Skip to Content
Merck
All Photos(1)

Documents

I0659

Sigma-Aldrich

Ingenol-3-angelate

≥98% (HPLC), amorphous

Synonym(s):

(1aR,2S,5R,5aS,6S,8aS,9R,10aR)-5,5a-Dihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-11-oxo-1a,2,5,5a,6,9,10,10a-octahydro-1H-2,8a-methanocyclopenta[a]cyclpropa[e][10]annulen-6-yl (2Z)-2-methylbut-2-enoate, 3-Angeloylingenol, 3-Ingenyl angelate, I3A, Ingenol mebutate, PEP005

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

amorphous

color

clear

solubility

DMSO: >1 mg/mL

storage temp.

−20°C

SMILES string

O=C1[C@@]2(C=C(C)[C@@H]3OC(/C(C)=C\C)=O)[C@]3(O)[C@H](O)C(CO)=C[C@@]1([H])[C@@](C4(C)C)([H])[C@@]4([H])C[C@H]2C

InChI

1S/C25H34O6/c1-7-12(2)22(29)31-21-13(3)10-24-14(4)8-17-18(23(17,5)6)16(20(24)28)9-15(11-26)19(27)25(21,24)30/h7,9-10,14,16-19,21,26-27,30H,8,11H2,1-6H3/b12-7-/t14-,16+,17-,18+,19-,21+,24+,25+/m1/s1

InChI key

VDJHFHXMUKFKET-WDUFCVPESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ingenol 3-Angelate is a diterpenoid ester isolated from Euphorbia peplus that induces rapid cell death with keratinocytes and some cancer cell lines. Ingenol 3-Angelate is a protein kinase C activator that displays potent antileukemic activity. Ingenol 3-Angelate (ingenol mebutate) is used for treatment of actinic keratosis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongcheol Cho et al.
The Journal of biological chemistry, 290(37), 22759-22770 (2015-07-15)
Microtubule dynamics are important for axon growth during development as well as axon regeneration after injury. We have previously identified HDAC5 as an injury-regulated tubulin deacetylase that functions at the injury site to promote axon regeneration. However, the mechanisms involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service