Skip to Content
Merck
All Photos(1)

Documents

I0274

Sigma-Aldrich

myo-Inositol Monophosphatase from bovine brain

lyophilized powder, 5-20 units/mg protein

Synonym(s):

myo-Inositol monophosphate phosphohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

5-20 units/mg protein

composition

Protein, ~0.5% Bradford

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

The enzyme plays a critical role in the recycling of inositol to phosphoinositides in the mammalian brain and participates in the phosphatidylinositol signaling pathway. It is inhibited non-competitively by Li+, which may be significant in the treatment of manic depression. The enzyme may help in identifying less toxic substitutes for lithium in manic depressive therapy.

Unit Definition

One unit will form 1.0 μmole of phosphate from inositol 1-phosphate per min at pH 7.4 at 37°C.

Physical form

Lyophilized powder containing HEPES buffer salts, pH 7.5, and stabilizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana Kalujnaia et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(10), 3981-3991 (2010-06-16)
This study investigated the expression and tissue distribution of inositol monophosphatase (IMPA1) and characterized its role in salinity adaptation in the eel. The coding sequence of eel IMPA1 was determined and confirmed to be orthologous to the mammalian gene/enzyme by
Christopher A Broberg et al.
Science (New York, N.Y.), 329(5999), 1660-1662 (2010-08-21)
The marine bacterium Vibrio parahaemolyticus causes gastroenteritis in humans and encodes the type III effector protein VPA0450, which contributes to host cell death caused by autophagy, cell rounding, and cell lysis. We found that VPA0450 is an inositol polyphosphate 5-phosphatase
S J Pollack et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(13), 5766-5770 (1994-06-21)
myo-Inositol monophosphatase (myo-inositol-1-phosphate phosphohydrolase, EC 3.1.3.25) is an attractive target for mechanistic investigation due to its critical role in the phosphatidylinositol signaling pathway and the possible relevance of its inhibition by Li+ to manic depression therapy. The x-ray crystallographic structure
Livia Pasquali et al.
Behavioural pharmacology, 21(5-6), 473-492 (2010-08-12)
This is a short overview focusing on the biochemical interactions underlying the protective effects of lithium at the neuronal level. These include lithium modulation of autophagy, growth factors, excitotoxicity, and a variety of mechanisms underlying cell death, neurogenesis, and neuronal
Toward a crystal-clear view of lithium's site of action.
J M Baraban
Proceedings of the National Academy of Sciences of the United States of America, 91(13), 5738-5739 (1994-06-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service