Skip to Content
Merck
All Photos(1)

Key Documents

HT254

Sigma-Aldrich

Van Gieson Solution

for in vitro diagnostic use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47
Pricing and availability is not currently available.

form

solution

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

Application

Intended for use in the Sigma Elastic Stain kit, HT25A. Stains collagen brilliant red and muscle and cytoplasm yellow.

Components

0.05% acid fuchsin in saturated picric acid.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Popowics et al.
Journal of biomechanics, 42(12), 1961-1966 (2009-06-09)
Alveolar bone supports teeth during chewing through a ligamentous interface with tooth roots. Although tooth loads are presumed to direct the development and adaptation of these tissues, strain distribution in the alveolar bone at different stages of tooth eruption and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service