Skip to Content
Merck
All Photos(2)

Key Documents

HS23243A

Wire Drying Rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$232.00

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EACH
$232.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

high-density polyethylene (coated steel wire)

feature

to hold up to 90 pieces of labware

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific HS23243A

L × W × H

462 mm × 182 mm × 525 mm

General description

The Wire Drying Rack saves glassware from chips and cracks. You can drain and dry test tubes, beakers, flasks, and other labware on the rack.

Features and Benefits

  • Place self-standing rack on a table or near a sink, or use hooks to hang rack on the wall
  • Holds up to 90 pieces of labware on 18 x 127 mm and 72 x 76 mm pins
  • Constructed of spot-welded, vinyl-coated steel wire for durability with a polystyrene tray and back
  • Easily accessible and removable bottom drainage pan

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service