Skip to Content
Merck
All Photos(1)

Documents

HS15991B

Slide box

100 place, red

Synonym(s):

Box for Slides, Slide Container, Slide Storage Box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123302
NACRES:
NB.17

material

polypropylene

feature

Holds 100 x slides 76x26 mm

manufacturer/tradename

Heathrow Scientific HS15991B

size

210x169x37 mm

color

red

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The slide box is made of durable polypropylene.
  • The box can hold either 25 or 100 standard slides (25 x 75 mm and 1 x 3 inches).
  • Latch locks keep hinged lids securely closed.
  • Stability is maximized by stacking feet.
  • Slide identification card - capacity 100 slides
  • No liner

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service