Select a Size
$138.00
Select a Size
About This Item
$138.00
Recommended Products
material
pink-purple
polycarbonate lid
polypropylene
feature
holds 96-Well, 0.2 mL and 0.5 mL Tubes, and Assay Plates
packaging
pkg of 2 ea
manufacturer/tradename
Heathrow Scientific 120727
L × W × H
110 mm × 148 mm × 48 mm
Looking for similar products? Visit Product Comparison Guide
General description
Specifications
- Modern patented design cools faster in freezers and stays cold longer on the bench top
- PCR Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
- When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
- Cools industry standard format 0.2 mL and 0.5 mL PCR tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
- Better alternative then ice baths as dry incubation reduces the risk of contamination
- Freeze cooler in an upright position enabling cold storage of tubes
- Well design prohibits the freezing of samples
- Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
- Save space with compact footprint and stacking ability
- The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
- Robust cooler made from engineered polypropylene and the lid is made from polycarbonate
- US Patent Pending, Community Design No: 004101756-0001-0002
- Two Year Warranty
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service