The length of this reservoir is 2.8 inches and is not suitable for a 12 channel pipette. Please see the link below to review multichannel pipette reservoirs that may be of interest:
https://www.sigmaaldrich.com/US/en/substance/texanreagentreservoirformultichannelpipettes1234598765
HS120638
Reagent Reservoir
capacity 10 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
Synonym(s):
Reagent container, Reagent storage
Select a Size
$94.60
Select a Size
About This Item
$94.60
Recommended Products
material
V-bottom bottom
polystyrene
sterility
non-sterile
packaging
pack of 100 (bulk packed)
manufacturer/tradename
Heathrow Scientific 120638
capacity
10 mL
General description
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
Does this reservoir fit a 12 multichannel pipette?
1 answer-
Helpful?
-