Key Documents
HS120114
Mega Rack® tube rack
double, for 13-16 mm tubes, white
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
Pricing and availability is not currently available.
material
polypropylene
feature
Holds 240 x 10 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120114
size
673x267x90 mm
color
white
General description
Mega Rack High-capacity storage rack for sample and blood collection tubes.
  • Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
  • Sturdy and light weight material for easy transport
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
  • Autoclavable
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)