Skip to Content
Merck
All Photos(2)

Key Documents

HPA068828

Sigma-Aldrich

Anti-FGFRL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

KATNGFGSLSVNYTLVVLDDISPGKESLGPDSSSGGQEDPASQQWARPRFTQPSKMRRRVIARPVGSSVRLKCVAS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGFRL1(53834)

General description

Fibroblast growth factor receptor (FGFR)-like protein 1 (FGFRL1) is a transmembrane receptor. It belongs to the FGFR family. FGFRL1 has three extracellular Ig-like domains and a single transmembrane domain and a short intracellular tail with a peculiar histidine-rich motif. It is located on human chromosome 4p16.

Immunogen

fibroblast growth factor receptor-like 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fibroblast growth factor receptor (FGFR)-like protein 1 (FGFRL1) has the ability to promote the fusion of CHO cells to multinucleated syncytia. It stimulates cell adhesion but not cell proliferation. FGFRL1 can interact with FGF ligands and heparin. It serves as a decoy receptor that impairs FGF signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST92821

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biology of FGFRL1, the fifth fibroblast growth factor receptor.
Trueb B.
Cellular and Molecular Life Sciences, 68(6), 951-964 (2011)
Examination of FGFRL1 as a candidate gene for diaphragmatic defects at chromosome 4p16. 3 shows that Fgfrl1 null mice have reduced expression of Tpm3, sarcomere genes and Lrtm1 in the diaphragm.
LopezJimenez N, et al.
Human Genetics, 127(3), 325-336 (2010)
Evolution of the fusogenic activity of the receptor FGFRL1.
Zhuang L and Trueb B
Archives of Biochemistry and Biophysics, 625, 54-64 (2017)
Receptor FGFRL1 does not promote cell proliferation but induces cell adhesion.
Yang X, et al.
International Journal of Molecular Medicine, 38(1), 30-38 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service