Skip to Content
Merck
All Photos(2)

Key Documents

HPA050121

Sigma-Aldrich

Anti-BST1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD157, Anti-bone marrow stromal cell antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

EGTSAHLRDIFLGRCAEYRALLSPEQRNKNCTAIWEAFKVALDKDPCSVLPSDYDLFINLSRHSIPRDKSLFWE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BST1(683)

Related Categories

General description

The gene BST1 (bone marrow stromal cell antigen 1) is mapped to human chromosome 4p15. It belongs to the CD38 (cluster of differentiation 38) gene family and the encoded protein is a member of the NADase (NAD+ glycohydrolase)/ADP (adenosine diphosphate)-ribosyl cyclase family.[1] The BST1 gene encodes a glycosylphosphatidylinositol-anchored protein. It is present in myeloid, endothelial and mesothelial cells as well as in epithelial ovarian cancer cells.

Immunogen

bone marrow stromal cell antigen 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BST1 (bone marrow stromal cell antigen 1) was recognized as a stromal and myeloid surface glycoprotein. It is responsible for the cleavage of extracellular nicotinamide adenine dinucleotide (NAD+), resulting in ADP ribose (ADPR) and cyclic ADPR (cADPR). BST1 helps in the transduction of intracellular signals by associating with transmembrane molecules. It is involved in leukocyte adhesion, migration and diapedesis.[1] Mutations in the BST1 gene are associated with susceptibility to Parkinson′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85380

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding of CD157 protein to fibronectin regulates cell adhesion and spreading.
Morone S, et al.
The Journal of Biological Chemistry, 289, 15588-15601 (2014)
Association between bone marrow stromal cell antigen 1 gene polymorphisms and the susceptibility to Parkinson's disease: a meta-analysis.
Wang S, et al.
Neuroscience Letters, 599, 120-124 (2015)
A deletion involving CD38 and BST1 results in a fusion transcript in a patient with autism and asthma.

A deletion involving CD38 and BST1 results in a fusion transcript in a patient with autism and asthma.
Ceroni F, et al.
Autism Research : Official Journal of the International Society for Autism Research, 7, 254-263 (2014)
Emiko Aomatsu et al.
Scientific reports, 4, 3652-3652 (2014-01-15)
Human mesenchymal stem cells (hMSCs) remodel or regenerate various tissues through several mechanisms. Here, we identified the hMSC-secreted protein SCRG1 and its receptor BST1 as a positive regulator of self-renewal, migration, and osteogenic differentiation. SCRG1 and BST1 gene expression decreased

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service