Skip to Content
Merck
All Photos(7)

Key Documents

HPA041656

Sigma-Aldrich

Anti-NUBP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NBP1, Anti-NBP35

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TSDEHLSVVRYLATAHIDGAVIITTPQEVSLQDVRKEINFCRKVKLPIIGVVEN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUBP1(4682)

General description

Nucleotide-binding protein 1 (NUBP1) is also called cytosolic Fe-S cluster assembly factor. The NUBP1 gene is mapped to human chromosome 16p13.3. It has a phosphate-binding loop (P-loop) motif and two highly conserved NUBP/MRP (multiple resistance and pH adaptation) motifs α and β.

Immunogen

nucleotide binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-NUBP1 antibody produced in rabbit has been used in the detection of NUBP1 in human HepG2 cells and HeLa cells using western blotting.

Biochem/physiol Actions

Nucleotide-binding protein 1 (NUBP1) plays an important role in the maturation of Fe/S proteins and their homeostasis. NUBP1 interacts with motor protein, kinesin family C-terminal 5A (KIFC5A) and modulates centrosome duplication. Nubp1 along with Nubp2, are associated with centrioles during cell cycle events. Duplications in the NUBP1 gene locus may contribute to intellectual disability and facial anomalies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82188

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two novel mouse genes?Nubp2, mapped to the t-complex on chromosome 17, and Nubp1, mapped to chromosome 16?establish a new gene family of nucleotide-binding proteins in eukaryotes
Nakashima H, et al.
Genomics, 60(2), 152-160 (1999)
Human Nbp35 is essential for both cytosolic iron-sulfur protein assembly and iron homeostasis
Stehling O, et al.
Molecular and Cellular Biology, 28(17), 5517-5528 (2008)
Motor protein KIFC5A interacts with Nubp1 and Nubp2, and is implicated in the regulation of centrosome duplication
Christodoulou A, et al.
Journal of Cell Science, 119(10), 2035-2047 (2006)
Dysfunction in the mitochondrial Fe-S assembly machinery leads to formation of the chemoresistant truncated VDAC1 isoform without HIF-1alpha activation
Ferecatu I, et al.
PLoS ONE, 13(3), e0194782-e0194782 (2018)
The diabetes drug target MitoNEET governs a novel trafficking pathway to rebuild an Fe-S cluster into cytosolic aconitase/iron regulatory protein 1
Ferecatu I, et al.
The Journal of Biological Chemistry, 28070?28086-28070?28086 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service