Skip to Content
Merck
All Photos(1)

Documents

HPA040361

Sigma-Aldrich

Anti-CX3CL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Abcd-3, Anti-C3xkine, Anti-Chemokine (c-x3-c motif) ligand 1, Anti-Cxc3, Anti-Cxc3c, Anti-Fractalkine, Anti-Neurotactin, Anti-Ntn, Anti-Scyd1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

GVTKCNITCSKMTSKIPVALLIHYQQNQASCGKRAIILETRQHRLFCADPKEQWVKDAMQHLDRQAAALTRNGGTFEKQIGEVKPRTTPAAGGMDESVVLE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CX3CL1(6376)

Related Categories

General description

C-X3-C motif chemokine ligand 1(CX3CL1), also known as fractalkine (FKN), is encoded by the gene mapped to human chromosome 16q21. The encoded protein acts as a chemotactic cytokine in a soluble form, or acts as a binding molecule in a membrane-attached form. CX3CL1 is characterized by a mucin-like stalk containing chemokine domain and single transmembrane domain with a short intracellular C-terminal. CX3CL1 belongs to the CX3C chemokine subfamily.

Immunogen

chemokine (C-X3-C motif) ligand 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-CX3CL1 antibody produced in rabbit has been used in tissue microarray (TMA) immunostaining.

Biochem/physiol Actions

CX3CL1 interacts with its cognate receptor CX3CR1 and stimulates chemotaxis of macrophages to apoptotic lymphocytes. The protein also facilitates inflammatory processes in the central nervous system (CNS). CX3CL1 has been implicated in the molecular mechanism that controls cell adhesion, migration and survival of human prostate cancer cells. Hence, the protein has potential therapeutic approach to treatment of prostate cancer. Elevated expression of serum CX3CL1 has been observed in postmenopausal osteoporotic patients. Thus, it acts as a potential diagnostic marker and therapeutic target for anti-resorptive treatment in osteoporosis patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81765

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucy A Truman et al.
Blood, 112(13), 5026-5036 (2008-09-19)
Cells undergoing apoptosis are efficiently located and engulfed by phagocytes. The mechanisms by which macrophages, the professional scavenging phagocytes of apoptotic cells, are attracted to sites of apoptosis are poorly defined. Here we show that CX3CL1/fractalkine, a chemokine and intercellular
Yi-Ding Chen et al.
British journal of biomedical science, 73(3), 121-128 (2016-08-02)
The chemokine (C-X3-C motif) ligand 1 (CX3CL1), also called fractalkine (FKN), has recently been reported to be involved in osteoclastogenic process and pathological bone destruction. This study aimed to investigate the link between serum CX3CL1/FKN levels with disease progression of
Gladys Morrison et al.
Stem cell research, 16(1), 140-148 (2016-01-17)
Differentiated cells retain the genetic information of the donor but the extent to which phenotypic differences between donors or batches of differentiated cells are explained by variation introduced during the differentiation process is not fully understood. In this study, we
WangMi Liu et al.
Archivum immunologiae et therapiae experimentalis, 64(5), 371-383 (2016-04-22)
Chemokines are a family of small 8-10 kDa inducible cytokines. Initially characterized as chemotactic factors, they are now considered to affect not just cellular recruitment. CX3CL1 is a unique chemokine that can exist in a soluble form, as a chemotactic cytokine
Jiebing Tang et al.
Oncology reports, 35(2), 1153-1162 (2016-01-01)
Epithelial-to-mesenchymal transition (EMT) endows cancer cells with enhanced invasive and metastatic potential during cancer progression. Fractalkine, also known as chemokine (C-X3-C motif) ligand 1 (CX3CL1), the only member recognized so far that belongs to the CX3C chemokine subfamily, was reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service