Skip to Content
Merck
All Photos(1)

Documents

HPA039832

Sigma-Aldrich

Anti-PTPRU antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fmi, Anti-Hptp-j, Anti-Pcp-2, Anti-Protein tyrosine phosphatase, receptor type, u, Anti-Ptp, Anti-Ptpro

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

TFEEASDPAVPCEYSQAQYDDFQWEQVRIHPGTRAPADLPHGSYLMVNTSQHAPGQRAHVIFQSLSENDTHCVQFSYFLYSRDG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRU(10076)

General description

Protein tyrosine phosphatase receptor type U (PTPRU) is also termed as purkinje cell protein 2 (PCP2)/ PTPλ. It belongs to the R2B subfamily of receptor protein tyrosine phosphatases (RPTPs). This gene is located on human chromosome 1p35.3.

Immunogen

protein tyrosine phosphatase, receptor type, U recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Protein tyrosine phosphatase receptor type U (PTPRU) negatively modulates the growth and migration of colon cancer cells. It participates in cell adhesion and acts as tumor suppressors. Knockdown of PTPRU helps to prevent the development of gastric cancer. In gastric cancer cells, PTPRU controls tyrosine phosphorylation and transcriptional activity of β-catenin. It is essential for glioma cell motility and in vivo glioma progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77703

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MicroRNA-574-5p promotes metastasis of non-small cell lung cancer by targeting PTPRU.
Zhou R, et al.
Scientific Reports, 6, 35714-35714 (2016)
Knockdown of protein tyrosine phosphatase receptor U inhibits growth and motility of gastric cancer cells.
Liu Y, et al.
International Journal of Clinical and Experimental Pathology, 7(9), 5750?5761-5750?5761 (2014)
Protein tyrosine phosphatases in the human genome.
Alonso A, et al.
Cell, 117(6), 699-711 (2004)
Zhichuan Zhu et al.
Carcinogenesis, 35(8), 1901-1910 (2014-05-31)
The membrane protein tyrosine phosphatase receptor U (PTPRU) has been shown to function as a negative regulator of adhesion and proliferation in certain cancer cell types, primarily through its dephosphorylation of β-catenin and inhibition of subsequent downstream signaling. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service