Skip to Content
Merck
All Photos(4)

Key Documents

HPA038736

Sigma-Aldrich

Anti-ANKRD1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALRP, Anti-C-193, Anti-CARP, Anti-CVARP, Anti-MCARP, Anti-ankyrin repeat domain 1 (cardiac muscle)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RYKMIRLLIMYGADLNIKNCAGKTPMDLVLHWQNGTKAIFDSLRENSYKTSRIATF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANKRD1(27063)

General description

The ankyrin repeat domain 1 (ANKRD1) gene mapped to human 10q23.33, encodes cardiac adriamycin responsive protein (CARP). The encoded nuclear protein is a member of the muscle ankyrin repeat proteins family. Members of this family contain four ankyrin repeats constituting a conserved motif. ANKRD1 is mainly expressed in heart during embryonic and fetal development.

Immunogen

ankyrin repeat domain 1 (cardiac muscle) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ankyrin repeat domain 1 (ANKRD1) might act as a transcriptional negative regulator for Ventricular myosin light chain-2 (MLC-2v) in cardiac myogenesis. The encoded protein plays a vital role in cardiac muscle function in both physiological and pathological conditions. Additionally, it is also implicated in the regulation of gene expression in the heart. Variations in the gene expression leads to hypertrophic cardiomyopathy (HCM) and human dilated cardiomyopathy. ANKRD1 functions as a transcriptional co-activator and enhances tumor suppressive activity of p53 protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80281

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping of porcine ANKRD1, ANKRD2, ANKRD23, VGLL2 and VGLL4 using somatic cell and radiation hybrid panels.
Dong XJ
Animal Genetics, 38, 424-425 (2007)
Assignment of the murine ankyrin-repeated protein gene (Ankrd2) to mouse chromosome 19C3-->D1 and rat chromosome 1q51-->q53 by fluorescence in situ hybridization.
Fujiwara M
Cytogenetic and genome research, 105 (2004)
Cardiac ankyrin repeat protein gene (ANKRD1) mutations in hypertrophic cardiomyopathy.
Arimura T
Journal of the American College of Cardiology, 54, 334-342 (2009)
A novel role for cardiac ankyrin repeat protein Ankrd1/CARP as a co-activator of the p53 tumor suppressor protein.
Kojic S
Archives of Biochemistry and Biophysics, 502, 60-67 (2010)
Mutations in the ANKRD1 gene encoding CARP are responsible for human dilated cardiomyopathy.
Duboscq-Bidot L
European Heart Journal, 30, 2128-2136 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service