Skip to Content
Merck
All Photos(5)

Documents

HPA035471

Sigma-Aldrich

Anti-MCAT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FASN2C, Anti-FabD, Anti-MCT, Anti-MT, Anti-Malonyl CoA:ACP acyltransferase (mitochondrial), Anti-NET62

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HKLLAQQLVSPVKWEQTMHAIYERKKGRGFPQTFEVGPGRQLGAILKSCNMQAWKSYSAVDVLQTLEHVDLDPQEP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCAT(27349)

General description

Malonyl-CoA-acyl carrier protein transacylase (MCAT) is encoded by the gene mapped to human chromosome 22q13.2. The encoded protein contains two subdomains, where one subdomain is composed of short four-stranded parallel β-sheet and 12 helices and the other subdomain contains a four-stranded antiparallel β-sheet and two helices.

Immunogen

malonyl CoA:ACP acyltransferase (mitochondrial) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Malonyl-CoA-acyl carrier protein transacylase (MCAT) is an important enzyme involved in the fatty-acid biosynthesis pathway of bacteria. This enzyme catalyzes the transfer of a malonyl group from malonyl–CoA to the free thiol group of the phosphopantetheine arm of acyl carrier protein. In addition, it is also implicated in polyketide biosynthesis, producing one of the largest classes of secondary metabolites, such as the tetracyclines and erythromycins. Hence, MCAT is considered as a key enzyme in bacterial metabolic activity. This enzyme also serves as a potential drug target in the discovery of antibacterial agents.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78177

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A continuous coupled enzyme assay for bacterial malonyl?CoA:acyl carrier protein transacylase (FabD)
Molnos J, et al.
Analytical Biochemistry, 319(1), 171-176 (2003)
Malonyl?CoA: acyl carrier protein transacylase from Helicobacter pylori: Crystal structure and its interaction with acyl carrier protein
Zhang L, et al.
Protein Science, 16(6), 1184-1192 (2007)
Prostate cancer risk-associated variants reported from genome-wide association studies: meta-analysis and their contribution to genetic variation
Kim S T, et al.
Prostate, 70(16), 1729-1738 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service