Skip to Content
Merck
All Photos(4)

Key Documents

HPA035428

Sigma-Aldrich

Anti-PCYT1A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CT, Anti-CTPCT, Anti-PCYT1, Anti-phosphate cytidylyltransferase 1, choline, α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DGVPSKVQRCAVGLRQPAPFSDEIEVDFSKPYVRVTMEEASRGTPCERPVR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCYT1A(5130)

General description

The gene PCYT1A (choline-phosphate cytidylyltransferase A) is mapped to human chromosome 3q29. It is widely expressed and works as a homodimer. The encoded protein has a nuclear localization signal sequence, a catalytic domain, a membrane binding domain (domain M) and an unstructured phosphorylated tail with up to 16 phosphoserine sites (domain P).

Immunogen

phosphate cytidylyltransferase 1, choline, alpha recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PCYT1A antibody produced in rabbit has been used in automated immunohistochemistry.

Biochem/physiol Actions

PCYT1A (choline-phosphate cytidylyltransferase A) is the rate limiting enzyme in the Kennedy phosphatidylcholine pathway. It is responsible for the generation of energy donor CDP (cytidine diphosphate)-choline. Mutations in this gene cause reduction in the phosphatidylcholine levels, thereby disrupting functioning of white adipose tissue and insulin activity. Mutations in this gene are also linked with spondylometaphyseal dysplasia with cone-rod dystrophy and isolated forms of retinal dystrophy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79426

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in the PCYT1A gene are responsible for isolated forms of retinal dystrophy.
Testa F
European Journal of Human Genetics, 25, 651-651 (2017)
Mutations disrupting the Kennedy phosphatidylcholine pathway in humans with congenital lipodystrophy and fatty liver disease.
Payne F
Proceedings of the National Academy of Sciences of the USA, 111, 8901-8901 (2014)
Tammer Hemdan et al.
Scandinavian journal of urology, 52(3), 200-205 (2018-02-25)
The aim of this study was to test choline-phosphate cytidylyltransferase-α (CCT-α) protein as a biomarker for neoadjuvant cisplatin chemotherapy response in a bladder tumor setting. A total of 238 patients with T2-T4 bladder cancer enrolled into two prior randomized trials
Choline-phosphate cytidylyltransferase-alpha as a possible predictor of survival and response to cisplatin neoadjuvant chemotherapy in urothelial cancer of the bladder
Hemdan T, et al.
Scandinavian journal of urology, 52(3), 200-205 (2018)
Mutations in PCYT1A, encoding a key regulator of phosphatidylcholine metabolism, cause spondylometaphyseal dysplasia with cone-rod dystrophy.
Hoover-Fong J
American Journal of Human Genetics, 94, 105-105 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service