Skip to Content
Merck
All Photos(8)

Key Documents

HPA035015

Sigma-Aldrich

Anti-STAG1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NYD-SP12, Anti-SA-1, Anti-stromal antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TSELPVLQDSTNETTAHSDAGSELEETEVKGKRKRGRPGRPPSTNKKPRKSPGEKSR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAG1(10274)

General description

Stromal antigen 1 (STAG1) is a nuclear protein and it is majorly expressed in hematopoietic organs and somatic cells. The gene is located on human chromosome 3q22.3. The protein belongs to the stromal antigen (STAG) subunit of core cohesin complex, that is regulated by PDS5B (PDS5 cohesin associated factor B).

Immunogen

stromal antigen 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-STAG1 antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

Stromal antigen 1 (STAG1) is an important mediator for p53-dependent apoptosis. It is critical for telomeric sister chromatid cohesion in mammalian cells. The protein contributes to a number of pivotal cellular processes, such as transcription, maintenance of chromatin domain architecture and insulator function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79351

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic lethal interaction between the tumour suppressor STAG2 and its paralog STAG1
Benedetti L, et al.
Oncotarget, 8(23), 37619-37619 (2017)
CTCF and cohesinSA-1 mark active promoters and boundaries of repressive chromatin domains in primary human erythroid cells
Steiner LA, et al.
PLoS ONE, 11(5), e0155378-e0155378 (2016)
Identification of STAG1 as a key mediator of a p53-dependent apoptotic pathway
Anazawa Y, et al.
Oncogene, 23(46), 7621-7621 (2004)
De novo gene mutations highlight patterns of genetic and neural complexity in schizophrenia
Xu B, et al.
Nature Genetics, 44(12), 1365-1365 (2012)
STAG1 mutations cause a novel cohesinopathy characterised by unspecific syndromic intellectual disability
Lehalle D, et al.
Journal of medical Genetics, jmedgenet-jmedg2016 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service