Skip to Content
Merck
All Photos(3)

Key Documents

HPA026794

Sigma-Aldrich

Anti-HOXC5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APT, Anti-HOX3, Anti-HOX3D, Anti-LYPLA2L, Anti-dJ570F3.6, Anti-homeobox C5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PGHAPGRDEAAPLNPGMYSQKAARPALEERAKSSGEIKEEQAQTGQPAGLSQPPAPPQIYPWMT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXC5(3222)

General description

Homeobox C5 (HOXC5), also known as HOX3D, is a member of HOX class I family. The gene coding for this protein is mapped to human chromosome 12. HOXC5 is expressed primarily in mature lymphoid cell lines and neoplastic cells of non-Hodgkin′s lymphomas (NHL). In normal lymphoid cells and leukemia′s, HOXC5 is not expressed.

Immunogen

homeobox C5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Homeobox C5 (HOXC5) is associated with the development of lymphomagenesis. The encoded protein might take part in process initiating, transformation of cervical keratinocytes. The A17860G positioned in 3′ flanking sequence of HOXC5 gene is involved in pathogenesis of simple congenital heart disease (CHD). HOXC5 might function as a potential biomarker and therapeutic target of oral squamous cell carcinoma (OSCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86523

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HOXC5 and HOXC8 expression are selectively turned on in human cervical cancer cells compared to normal keratinocytes.
Alami Y
Biochemical and Biophysical Research Communications, 257(3), 738-745 (1999)
Li-guo Gong et al.
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics, 22(5), 497-501 (2005-10-11)
In the candidate region 12q13 of simple congenital heart disease(CHD), four single nucleotide polymorphisms(SNPs) in HOXC4 gene were chosen in order to investigate the distribution of SNP and haplotypes in simple CHD patients and normal people. The genotype of 4
Chung-Jung Li et al.
Nature communications, 8, 14685-14685 (2017-03-25)
The initial rostrocaudal patterning of the neural tube leads to differential expression of Hox genes that contribute to the specification of motor neuron (MN) subtype identity. Although several 3' Hox mRNAs are expressed in progenitors in a noisy manner, these
J Bijl et al.
Blood, 87(5), 1737-1745 (1996-03-01)
Besides their regulatory role in embryogenesis, homeobox (HOX) genes are expressed in a specific manner in hematopoietic cell lineages, implying a role in the molecular regulation of hematopoiesis. Some HOX C cluster genes are found to be expressed in lymphoid
Sung-Min Moon et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 41(6), 470-476 (2012-03-06)
Aberrant expression of homeobox genes (HOX), normally required for the differentiation of a particular tissue, has been reported in several types of cancer, but poorly addressed in oral squamous cell carcinoma (OSCC). The present study investigated the expression of HOXC5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service