Skip to Content
Merck
All Photos(3)

Key Documents

HPA026078

Sigma-Aldrich

Anti-ARFGEF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Brefeldin A-inhibited GEP 2, Anti-Brefeldin A-inhibited guanine nucleotide-exchange protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KPIQSKPQSPVIQAAAVSPKFVRLKHSQAQSKPTTPEKTDLTNGEHARSDSGKVSTENGDAPRERGSSLSGTDDGAQEVVKDILEDVVTSAIKAAEKHGLTEPERVLGELECQECAIPPGVDENSQTNGI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARFGEF2(10564)

General description

ADP ribosylation factor guanine nucleotide exchange factor 2 (ARFGEF2) possesses a Sec7 domain which is crucial for its activity. This protein is expressed in the brain and the gene encoding it is localized on human chromosome 20.

Immunogen

Brefeldin A-inhibited guanine nucleotide-exchange protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. The antibodies are also tested using immunofluorescence. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ADP ribosylation factor guanine nucleotide exchange factor 2 (ARFGEF2) activates ADP-ribosylation factors (ARFs) by exchanging guanosine diphosphate (GDP) with guanosine triphosphate (GTP). It aids in the release of type I tumor necrosis factor receptor (TNFR1) from human umbilical vein endothelial cells. Mutation in the ARFGEF2 gene has been linked to microcephaly and periventricular heterotopia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71270

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Y de Wit et al.
Neurogenetics, 10(4), 333-336 (2009-04-23)
We report a child with a severe choreadystonic movement disorder, bilateral periventricular nodular heterotopia (BPNH), and secondary microcephaly based on compound heterozygosity for two new ARFGEF2 mutations (c.2031_2038dup and c.3798_3802del), changing the limited knowledge about the phenotype. The brain MRI
Xiaoyan Shen et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(36), 14464-14469 (2012-08-22)
Brefeldin A-inhibited guanine nucleotide-exchange protein (BIG)2 activates ADP-ribosylation factors, ∼20-kDa GTPase proteins critical for continuity of intracellular vesicular trafficking by accelerating the replacement of ADP-ribosylation factor-bound GDP with GTP. Mechanisms of additional BIG2 function(s) are less clear. Here, the participation
Chantal Christis et al.
The Journal of cell biology, 196(3), 327-335 (2012-02-01)
The small G protein Arf1 regulates Golgi traffic and is activated by two related types of guanine nucleotide exchange factor (GEF). GBF1 acts at the cis-Golgi, whereas BIG1 and its close paralog BIG2 act at the trans-Golgi. Peripheral membrane proteins
Ehud Banne et al.
Journal of medical genetics, 50(11), 772-775 (2013-07-03)
West syndrome (WS) is an epileptic encephalopathy of childhood, defined by the presence of clustered spasms usually occurring before the age of 1 year, hypsarrhythmia on EEG that is notoriously difficult to define, and developmental arrest or regression. The incidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service