Skip to Content
Merck
All Photos(6)

Key Documents

HPA025703

Sigma-Aldrich

Anti-BLZF1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Basic leucine zipper nuclear factor 1, Anti-Golgin-45, Anti-JEM-1, Anti-p45 basic leucine-zipper nuclear factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KSLGHHKGEFLGQSEGVIEPNKELSEVKNVLEKLKNSERRLLQDKEGLSNQLRVQTEVNRELKKLLVASVGDDLQYHFERLARE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BLZF1(8548)

General description

The gene BLZF1 (basic leucine zipper nuclear factor 1) is mapped to human chromosome 1q24. It belongs to the b-ZIP family of proteins. The encoded protein has a PEST (proline [P], glutamic acid [E], serine [S], and threonine [T]) motif, a putative leucine repeat and several phosphorylation sites. This protein is present in the nucleus, cytoplasm and Golgi apparatus.

Immunogen

Golgin-45 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BLZF1 (basic leucine zipper nuclear factor 1) might work as a transcription factor. It is involved in epigenetic regulation by binding to DNMT3L (DNA methyltransferase 3 like) and also participates in Wnt/β-catenin signaling. The BLZF1 gene is downregulated in acute promyelocytic leukemia (APL) cell line NB4. On the other hand, it is upregulated in presence of dengue virus infection. BLZF1 also interacts with GRASP55 (the medial-Golgi matrix protein) and the GTP form of rab2. It is involved in the secretory protein transport and maintenance of the Golgi structure. This gene is downregulated in hepatocellular carcinoma. This low expression is associated with poor overall survival.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76646

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Short et al.
The Journal of cell biology, 155(6), 877-883 (2001-12-12)
Membrane traffic between the endoplasmic reticulum (ER) and Golgi apparatus and through the Golgi apparatus is a highly regulated process controlled by members of the rab GTPase family. The GTP form of rab1 regulates ER to Golgi transport by interaction
Run-Yue Huang et al.
Biochemical and biophysical research communications, 467(3), 602-609 (2015-09-08)
BLZF1, a member of b-ZIP family, has been implicated in epigenetic regulation and Wnt/β-catenin signaling. Its expression and clinical significance in human cancers remain largely unknown. In this study, we showed that BLZF1 expression was reduced in hepatocellular carcinoma (HCC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service