Skip to Content
Merck
All Photos(8)

Key Documents

HPA024705

Sigma-Aldrich

Anti-VPS37A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Anti-FLJ32642, Anti-HCRP1, Anti-PQBP2, Anti-SPG53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DVPDAFPELSELSVSQLTDMNEQEEVLLEQFLTLPQLKQIITDKDDLVKSIEELARKNLLLEPSLEAKRQTVLDKYELLTQMKSTF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VPS37A(137492)

General description

The gene VPS37A (vacuolar protein sorting-associated protein 37A) is mapped to human chromosome 8p22. It is part of the ESCRT-I (endosomal complexes required for transport) complex.

Immunogen

vacuolar protein sorting 37 homolog A (S. cerevisiae) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VPS37A (vacuolar protein sorting-associated protein 37A) is a part of the ESCRT-I (endosomal complexes required for transport) complex, which participates in membrane remodelling, including multivesicular body biogenesis and cytokinesis. VPS37A is downregulated in various cancers, including ovarian cancer, hepatocellular carcinoma, oral and oropharyngeal squamous cell carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76304

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Perisanidis et al.
Oral diseases, 19(2), 206-211 (2012-08-16)
The hepatocellular carcinoma-related protein 1 (HCRP1) is a key factor in the degradation of the epidermal growth factor receptor. In this study, we assessed the prognostic significance of HCRP1 expression in patients with oral and oropharyngeal squamous cell carcinoma (OOSCC).
Zhenhua Xu et al.
Biochemical and biophysical research communications, 311(4), 1057-1066 (2003-11-19)
One of the most frequent allelic deletions in hepatocellular carcinoma (HCC) has been found at chromosome 8p21-23. We reported here the identification and characterization of a novel gene for a hepatocellular carcinoma related protein 1 (HCRP1) localized at 8p22, which
Michael Wittinger et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(24), 7816-7827 (2011-10-22)
Although prognostic and predictive factors in ovarian cancer have been extensively studied for decades, only few have been identified and introduced to clinical practice. Here, we evaluate hVps37A (HCRP1) as a possible novel predictive marker for ovarian cancer. hVps37A was
Emily A Bruce et al.
Virology, 390(2), 268-278 (2009-06-16)
The mechanism of membrane scission during influenza A virus budding has been the subject of controversy. We confirm that influenza M1 binds VPS28, a subunit of the ESCRT-1 complex. However, confocal microscopy of infected cells showed no marked colocalisation between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service