Skip to Content
Merck
All Photos(6)

Documents

HPA024629

Sigma-Aldrich

Anti-KLF17 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Krueppel-like factor 17, Anti-Zinc finger protein 393

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TVPSTEAQAVLPSMAQMLPPQDAHDLGMPPAESQSLLVLGSQDSLVSQPDSQEGPFLPEQPGPAPQTVEKNSRPQEGTG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF17(128209)

General description

The gene KLF17 (Kruppel like factor 17) is mapped to human chromosome 1p34. The protein has a conserved DNA binding domain (DBD) containing C2H2 Kruppel like zinc fingers. It belongs to the Sp/KLF (specificity protein/Kruppel like factor) zinc finger protein family.

Immunogen

Krueppel-like factor 17 recombinant protein epitope signature tag (PrEST)

Application

Anti-KLF17 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

KLF17 (Kruppel like factor 17) interacts with the G/C and CACCC boxes of specific genes and is involved in transcription. KLF17 inhibits cancer cell metastasis, by suppressing the transcription of ID1 (inhibitor of DNA binding 1). It is downregulated in breast cancer, lung adenocarcinoma and hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76042

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William A Pastor et al.
Nature cell biology, 20(5), 553-564 (2018-04-27)
Naive and primed pluripotent human embryonic stem cells bear transcriptional similarity to pre- and post-implantation epiblast and thus constitute a developmental model for understanding the pluripotent stages in human embryo development. To identify new transcription factors that differentially regulate the
Paul Blakeley et al.
Development (Cambridge, England), 142(20), 3613-3613 (2015-10-22)
There were errors published in Development 142, 3151-3165.In the issue published online on 22 September 2015, Fig. 3 was mislabelled: panels A, B, C and D should have been B, C, D and A, respectively. In the legend, the text
Zhipeng Ai et al.
Cell reports, 40(8), 111240-111240 (2022-08-25)
Endogenous retroviruses (ERVs) have been reported to participate in pre-implantation development of mammalian embryos. In early human embryogenesis, different ERV sub-families are activated in a highly stage-specific manner. How the specificity of ERV activation is achieved remains largely unknown. Here
Bruno Di Stefano et al.
Nature methods, 15(9), 732-740 (2018-08-22)
Human embryonic stem cells (hESCs) can be captured in a primed state in which they resemble the postimplantation epiblast, or in a naive state where they resemble the preimplantation epiblast. Naive-cell-specific culture conditions allow the study of preimplantation development ex
Jane van Vliet et al.
Genomics, 87(4), 474-482 (2006-02-08)
The Sp/KLF transcription factors perform a variety of biological functions, but are related in that they bind GC-box and CACCC-box sequences in DNA via a highly conserved DNA-binding domain. A database homology search, using the zinc finger DNA-binding domain characteristic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service