Skip to Content
Merck
All Photos(4)

Documents

HPA023890

Sigma-Aldrich

Anti-CMTM4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CKLF-like MARVEL transmembrane domain-containing protein 4, Anti-Chemokine-like factor superfamily member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GEELDGFEGEASSTSMISGASSPYQPTTEPVSQRRGLAGLRCDPDYL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CMTM4(146223)

General description

CKLF (chemokine-like factor) like MARVEL transmembrane domain containing 4 (CMTM4) belongs to the CKLF like MARVEL transmembrane domain containing family (CMTM). The gene is located on human chromosome 16q21-q22.1. It is highly expressed in kidney, heart and brain.

Immunogen

CKLF-like MARVEL transmembrane domain-containing protein 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CKLF (chemokine-like factor) like MARVEL transmembrane domain containing 4 (CMTM4) functions as a regulator of cell cycle and division in HeLa cells. It acts as a tumor suppressor in clear cell renal cell carcinoma, when downregulated. The protein is involved in cell cycle growth. It regulates the protein programmed death-ligand 1 (PD-L1). It acts a target for microRNA-205 and enhances renal cell apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73177

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CMTM4 is frequently downregulated and functions as a tumour suppressor in clear cell renal cell carcinoma
Li T, et al.
Journal of Experimental & Clinical Cancer Research, 34(1), 122-122 (2015)
MicroRNA-205 inhibits renal cells apoptosis via targeting CMTM4
Zhang H, et al.
Iranian Journal of Basic Medical Sciences, 18(10), 1020-1020 (2015)
Integrated analysis of copy number and loss of heterozygosity in primary breast carcinomas using high-density SNP array
Ching HC, et al.
International Journal of Oncology, 39(3), 621-633 (2011)
Identification of CMTM6 and CMTM4 as PD-L1 protein regulators
Mezzadra R, et al.
Nature, 549(7670), 106-106 (2017)
Markus Plate et al.
Molecules and cells, 29(4), 355-361 (2010-03-10)
Human CMTM is a novel gene family consisting of CKLF and CMTM1-8. CMTM4 is the most conserved gene and has three RNA splicing forms designated as CMTM4-v1, -v2 and -v3, but in many types of tissue and cell lines, only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service