Skip to Content
Merck
All Photos(3)

Key Documents

HPA023599

Sigma-Aldrich

Anti-ZNF160 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HKr18, Anti-HZF5, Anti-Zinc finger protein 160, Anti-Zinc finger protein 5, Anti-Zinc finger protein Kr18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

LVSLGLCHFDMNIISMLEEGKEPWTVKSCVKIARKPRTRECVKGVVTDIPPKCTIKDLLPKEKSSTEAVFHTVVLERHESPDIEDFSFKEPQKNVHDFECQWRDDTGNYKGVLMAQKEGKRDQRDRRDIENKLMNNQLGVSFHSHLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF160(90338)

General description

The gene ZNF160 (zinc finger protein 160) is mapped to human chromosome 19q13.3-q13.4. It encodes a KRAB (Krüppel associated box) zinc finger protein with a molar mass of 94kDa containing 20 zinc finger motifs in its C terminus. The N terminus contains KRAB A and KRAB B domains. It is ubiquitously expressed.

Immunogen

Zinc finger protein 160 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene ZNF160 (zinc finger protein 160) encodes a transcriptional regulator that recruits histone deacetylase by interaction with KAP1 (KRAB [Kruppel-Associated Box Domain]-Associated Protein 1), a scaffold protein. Histone deacetylation and DNA methylation at the 5′ of the TLR4 (toll-like receptor) gene cause repression of this gene, possibly contributing to the maintenance of intestinal homeostasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83537

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular cloning and preliminary functional analysis of two novel human KRAB zinc finger proteins, HKr18 and HKr19.
Mark C
Dna and Cell Biology, 20, 275-286 (2001)
A novel C2H2 zinc-finger protein gene (ZNF160) maps to human chromosome 19q13.3-q13.4.
Halford S
Genomics, 25, 322-323 (1995)
Structure and mutagenesis of the Dbl homology domain.
Aghazadeh B
Nature Structural Biology, 5, 1098-1107 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service