Skip to Content
Merck
All Photos(3)

Key Documents

HPA023564

Sigma-Aldrich

Anti-CTBP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-C-terminal-binding protein 2, Anti-CtBP2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VPSYGVLGSRTSWDPMQGRSPALQDAGHLYRDPGGKMIPQGRQTQSRAASPGRYGREQPDTRYGAEVPAYPLSQVFSDISERPIDPAP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTBP2(1488)

General description

The gene CTBP2 (C-terminal binding protein 2) encodes a member of the CtBP-family. The gene is mapped to human chromosome 21q21.3. It is found to be expressed ubiquitously, with higher expression in the heart, skeletal muscle, and pancreas.

Immunogen

C-terminal-binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene CTBP2 (C-terminal binding protein 2) encodes a protein that functions as a transcriptional co-repressor of several tumor suppressor genes resulting in enhanced cancer cell migration and invasion. Its expression is found to be upregulated in hepatocellular carcinoma (HCC). It may be a potential prognostic marker for post liver resection HCC. It is involved in several types of tumor initiation, progression and response to therapy. It is found to interact with the C-terminal region of adenovirus type 2/5 E1A protein, a region that negatively regulates tumorigenicity and the extent of oncogenic transformation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75501

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zheng et al.
Oncotarget, 6(6), 3752-3769 (2015-02-18)
C-terminal binding protein 2 (CtBP2) is a transcriptional co-repressor that promotes cancer cell migration and invasion by inhibiting multiple tumor suppressor genes that contribute to cell mobility and adhesion. In this investigation, we showed thatCtBP2 expression was increased significantly in
CtBP2 could promote prostate cancer cell proliferation through c-Myc signaling.
Zhang C
Gene, 546, 73-79 (2014)
A novel C-terminal binding protein (CTBP2) is closely related to CTBP1, an adenovirus E1A-binding protein, and maps to human chromosome 21q21.3.
Katsanis N and Fisher EM
Genomics, 47, 294-299 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service