Skip to Content
Merck
All Photos(2)

Key Documents

HPA023331

Sigma-Aldrich

Anti-ZDHHC2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ZNF372, Anti-zinc finger, DHHC-type containing 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

KEFHLSYAEKDLLEREPRGEAHQEVLRRAAKDLPIYTRTMSGAIRYCDRCQLI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZDHHC2(51201)

General description

The gene ZDHHC2 (zinc finger DHHC-type containing 2) is mapped to human chromosome 8p21-22. It belongs to the palmitoyl acyltransferase (PAT) family and contains an Asp-His-His-Cys (DHHC) motif. PATs are mainly responsible for protein palmitoylation.[1] ZDHHC2 is present in mobile vesicles of dendrites.
Zinc finger DHHC-type containing 2 (ZDHHC2) is a tumor suppressor gene.

Immunogen

zinc finger, DHHC-type containing 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZDHHC2 (zinc finger DHHC-type containing 2) interacts with PSD95 (postsynaptic density protein 95), SNAP25 (synaptosomal-associated protein 25), SNAP23, CKAP4 (cytoskeleton-associated protein 4), CD9 and CD151. It plays a crucial role in the development, metastasis as well as recurrence of many cancers.[1] ZDHHC2 is suggested as a tumor/metastasis suppressor gene.
Zinc finger DHHC-type containing 2 (ZDHHC2) mediates palmitoylation of cytoskeleton-associated protein 4 (CKAP4), which is essential for CKAP4 subcellular division and antiproliferative factor (APF) stimulated cellular proliferation and gene expression. The protein plays an important role in metastasis and recurrence in human hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76196

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palmitoylation of cytoskeleton associated protein 4 by DHHC2 regulates antiproliferative factor-mediated signaling
Planey SL, et al.
Molecular Biology of the Cell, 20(5), 1454-1463 (2009)
A critical role for ZDHHC2 in metastasis and recurrence in human hepatocellular carcinoma
Peng C, et al.
BioMed Research International, 2014, e56366-e56366 (2014)
Chuanhui Peng et al.
BioMed research international, 2014, 832712-832712 (2014-07-06)
It has been demonstrated that loss of heterozygosity (LOH) was frequently observed on chromosomes 8p22-p23 in hepatocellular carcinoma (HCC) and was associated with metastasis and prognosis of HCC. However, putative genes functioning on this chromosomal region remain unknown. In this
Shu-Mei Yan et al.
PloS one, 8(2), e56366-e56366 (2013-03-05)
Zinc finger, DHHC-type containing 2 (ZDHHC2), originally named as reduced expression associated with metastasis protein (REAM), has been proposed as a putative tumor/metastasis suppressor gene and is often aberrantly decreased in human cancers. However ZDHHC2 expression pattern and its clinical
Jennifer Greaves et al.
Molecular biology of the cell, 22(11), 1887-1895 (2011-04-08)
Intracellular palmitoylation dynamics are regulated by a large family of DHHC (Asp-His-His-Cys) palmitoyl transferases. The majority of DHHC proteins associate with endoplasmic reticulum (ER) or Golgi membranes, but an interesting exception is DHHC2, which localizes to dendritic vesicles of unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service