Skip to Content
Merck
All Photos(5)

Documents

HPA023295

Sigma-Aldrich

Anti-NPLOC4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Nuclear protein localization protein 4 homolog, Anti-Protein NPL4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LEPFDEDYLNHLEPPVKHMSFHAYIRKLTGGADKGKFVALENISCKIKSGCEGHLPWPNGICTKCQPSAITLNRQKYRHVDNIMFENHTVADRFLDFWRKTGNQHFGYL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPLOC4(55666)

General description

The gene NPLOC4 (nuclear protein localization protein 4 homolog) is mapped to human chromosome 17q. It is mainly expressed in the heart, brain, kidney and skeletal muscle.

Immunogen

Nuclear protein localization protein 4 homolog recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The p97-NPLOC4 (nuclear protein localization protein 4 homolog)-UFD1 (ubiquitin fusion degradation 1 like) complex is responsible for the partial degradation of p100 and thereby positively controls the alternative nuclear factor κB pathway. The complex also plays an important role in the cell cycle regulation and DNA damage response. It inhibits chromosome instability by maintaining G2/M checkpoint signaling. NPLOC4 interacts with NEDD8 (neural precursor cell expressed, developmentally down-regulated 8) and is responsible for its degradation via p97-NPLOC4-UFD1 complex. VCP (valosin containing protein)-NPLOC4-UFD1 complex is important for endoplasmic reticulum-related degradation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75900

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Liu et al.
The Journal of biological chemistry, 288(43), 31339-31349 (2013-09-11)
The NEDD8 protein and neddylation levels in cells are modulated by NUB1L or NUB1 through proteasomal degradation, but the underlying molecular mechanism is not well understood. Here, we report that NUB1L down-regulated the protein levels of NEDD8 and neddylation through
Anne Riemer et al.
Cell cycle (Georgetown, Tex.), 13(6), 919-927 (2014-01-17)
The p97-Ufd1-Npl4 ATPase complex is associated with the response to DNA damage and replication stress, but how its inactivation leads to manifestation of chromosome instability is unclear. Here, we show that p97-Ufd1-Npl4 has an additional direct role in the G2/M
Zhao Zhang et al.
The Journal of biological chemistry, 290(32), 19558-19568 (2015-06-27)
Partial degradation of the p100 subunit to generate p52 subunit is a hallmark of the alternative NF-κB pathway, which has been implicated in cancer. Here, we uncovered a role of the p97-Npl4-Ufd1 complex in mediating p100-to-p52 processing and therefore positively
A Botta et al.
Gene, 275(1), 39-46 (2001-09-28)
The ubiquitin fusion-degradation gene (UFD1L) encodes the human homologue of the yeast ubiquitin fusion-degradation 1 protein, an essential component of the ubiquitin-dependent proteolytic turnover and mRNA processing. Although the UFD1L gene has been mapped in the region commonly deleted in
Jae-Jin Lee et al.
The Journal of biological chemistry, 288(10), 6998-7011 (2013-01-08)
Fas-associated factor 1 (FAF1) is a ubiquitin receptor containing multiple ubiquitin-related domains including ubiquitin-associated (UBA), ubiquitin-like (UBL) 1, UBL2, and ubiquitin regulatory X (UBX). We previously showed that N-terminal UBA domain recognizes Lys(48)-ubiquitin linkage to recruit polyubiquitinated proteins and that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service