Skip to Content
Merck
All Photos(3)

Key Documents

HPA021677

Sigma-Aldrich

Anti-PLCD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1, Anti-PLC-III, Anti-PLC-delta-1, Anti-Phosphoinositide phospholipase C, Anti-Phospholipase C-delta-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

QDDEDLQALLKGSQLLKVKSSSWRRERFYKLQEDCKTIWQESRKVMRTPESQLFSIEDIQEVRMGHRTEGLEKFARDVPEDRCFSIVFKDQRNTLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLCD1(5333)

General description

Phospholipase C-δ1 (PLCD1) is located in chromosome 3p21.3-p22 in humans. It is expressed in all normal adult tissues and is abundantly present in human nail matrix. Its functional attribute is highly dependent on intracellular free calcium concentration.

Immunogen

1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Phospholipase C- δ1 (PLCD1) is involved in the regulation of energy metabolism, calcium homeostasis and intracellular movement. It plays a crucial role in the phosphoinositide metabolism by hydrolyzing phosphatidylinositol-4, 5-bisphosphate to inositol 1, 4, 5 trisphosphate (IP3) and diacylglycerol. It has been studied that PLCD1 also possess tumor suppressive property in gastric cancer and breast cancer. Mutations in PLCD1 gene have been linked to a rare nail disorder, hereditary leukonychia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74720

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maija Kiuru et al.
American journal of human genetics, 88(6), 839-844 (2011-06-15)
Hereditary leukonychia (porcelain nails or white nails) is a rare nail disorder with an unknown genetic basis. To identify variants in a gene underlying this phenotype, we identified four families of Pakistani origin showing features of hereditary leukonychia. All 20
Shuji Shibutani et al.
Circulation, 125(8), 1027-1036 (2012-01-24)
We reported that phospholipase C (PLC)-δ1 activity was enhanced 3-fold in patients with coronary spastic angina. We detected variant PLC-δ1 with replacement of arginine 257 by histidine (R257H) showing increased enzymatic activity. We tested the hypothesis that increased PLC-δ1 activity
X-T Hu et al.
Oncogene, 28(26), 2466-2475 (2009-05-19)
Located at the important tumor suppressor locus, 3p22, PLCD1 encodes an enzyme that mediates regulatory signaling of energy metabolism, calcium homeostasis and intracellular movements. We identified PLCD1 as a downregulated gene in aerodigestive carcinomas through expression profiling and epigenetic characterization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service