Skip to Content
Merck
All Photos(4)

Key Documents

HPA020659

Sigma-Aldrich

Anti-SLCO2B1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-OATP-B, Anti-OATP-RP2, Anti-OATPRP2, Anti-Organic anion transporter B, Anti-Organic anion transporter polypeptide-related protein 2, Anti-Solute carrier family 21 member 9, Anti-Solute carrier organic anion transporter family member 2B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

GITHQTSAHPGLELSPSCMEACSCPLDGFNPVCDPSTRVEYITPCHAGCSSWVVQDALDNSQVFYTNCSCVVEGNPVLAGSCDSTCSHLVVPF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLCO2B1(11309)

General description

Solute carrier organic anion transporter family member 2B1 (SLCO2B1) is widely expressed. It is present in liver, placenta, small intestine, kidney, brain, skin, heart, platelets, and skeletal muscle.

Immunogen

Solute carrier organic anion transporter family member 2B1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier organic anion transporter family member 2B1 (SLCO2B1) is a transporter of various compounds. In the liver, it is involved in the uptake of amphiphilic organic anions.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86076

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyoung-Ah Kim et al.
Journal of clinical pharmacology, 53(11), 1186-1193 (2013-08-24)
Montelukast, a leukotriene receptor antagonist, is a substrate of organic anion transporting OATP2B1 encoded by the SLCO2B1. We evaluated the effects of six non-synonymous (c.1175C>T, c.1457C>T, c.43C>T, c.935G>A, c.601G>A, and c.644A>T) polymorphisms and one promoter (g.-282G>A) polymorphism on the pharmacokinetics
Michael J Knauer et al.
Molecular pharmacology, 83(6), 1218-1228 (2013-03-28)
Human organic anion transporting polypeptide 2B1 (OATP2B1) is a membrane transporter that facilitates the cellular uptake of a number of endogenous compounds and drugs. OATP2B1 is widely expressed in tissues including the small intestine, liver, kidney, placenta, heart, skeletal muscle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service