Skip to Content
Merck
All Photos(9)

Key Documents

HPA019948

Sigma-Aldrich

Anti-RUVBL1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-49 kDa TATA box-binding protein-interacting protein, Anti-49 kDa TBP-interacting protein, Anti-ECP-54, Anti-INO80 complex subunit H, Anti-NMP 238, Anti-Pontin 52, Anti-RuvB-like 1, Anti-TAP54-alpha, Anti-TIP49a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

GPPGTGKTALALAIAQELGSKVPFCPMVGSEVYSTEIKKTEVLMENFRRAIGLRIKETKEVYEGEVTELTPCETENPMGG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RUVBL1(8607)

General description

RUVBL1 (RuvB-like AAA ATPase 1) is a 50kDa protein belonging to the AAA(+)-family of ATPases (ATPase associated with diverse cellular activities). It is composed of three functional domains, named as (DI, DII, and DIII domains. DII domain consists of 6 α-helices, 9 β-strands, and 2 very short 310-helices. Among all the domains, two domains are responsible for ATP binding and hydrolysis.

Immunogen

RuvB-like 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RUVBL1 (RuvB-like AAA ATPase 1) is involved in various cellular processes including transcriptional regulation, DNA replication, DNA damage repair, chromatin remodeling and apoptosis. In the chromatin-remodeling process, it conjugates with other proteins to form the human histone acetylase/chromatin-remodeling complex TIP60, which plays an essential role in transcription and DNA repair. The chromatin-remodeling complex regulates chromatin structure as well as it maintains DNA-based export in the cell. It also functions as a component of the human RNA polymerase II holoenzyme complex, which indicates its role in transcriptional processes. RUVBL1 is also associated with two oncogenic pathways, c-Myc and another, β-catenin pathways. In addition, it also performs in small nucleolar ribonucleoprotein particle assembly, nucleolar localization, and trafficking.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74003

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justyna Durślewicz et al.
Discover. Oncology, 13(1), 106-106 (2022-10-16)
The present study aimed to investigate expression levels and prognostic significance of RUVBL1 and HNRNPU in stage I and II non-small-cell lung cancer (NSCLC) patients. Therefore, we evaluated immunohistochemical staining of RUVBL1 and HNRNPU, as well as RNA-seq data from
Justyna Durślewicz et al.
Cancers, 16(7) (2024-04-13)
This study investigates the roles of RUVBL1 and HIF1A in ccRCC development and explores their clinical significance as prognostic biomarkers. mRNA and protein expressions were analyzed using TCGA data and an institutional tissue cohort, respectively. Correlations with clinicopathological parameters and
Pedro M Matias et al.
The Journal of biological chemistry, 281(50), 38918-38929 (2006-10-25)
RuvBL1 is an evolutionarily highly conserved eukaryotic protein belonging to the AAA(+)-family of ATPases (ATPase associated with diverse cellular activities). It plays important roles in essential signaling pathways such as the c-Myc and Wnt pathways in chromatin remodeling, transcriptional and
Matteo Da Ros et al.
Autophagy, 13(2), 302-321 (2016-12-09)
Ribonucleoprotein (RNP) granules play a major role in compartmentalizing cytoplasmic RNA regulation. Haploid round spermatids that have exceptionally diverse transcriptomes are characterized by a unique germ cell-specific RNP granule, the chromatoid body (CB). The CB shares many characteristics with somatic
Jirina Tyleckova et al.
International journal of molecular sciences, 13(12), 15536-15564 (2013-02-28)
A comprehensive proteome map of T-lymphoblastic leukemia cells and its alterations after daunorubicin, doxorubicin and mitoxantrone treatments was monitored and evaluated either by paired comparison with relevant untreated control and using multivariate classification of treated and untreated samples. With the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service