Skip to Content
Merck
All Photos(6)

Key Documents

HPA019880

Sigma-Aldrich

Anti-CBLB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Casitas B-lineage lymphoma proto-oncogene b, Anti-E3 ubiquitin-protein ligase CBL-B, Anti-RING finger protein 56, Anti-SH3-binding protein CBL-B, Anti-Signal transduction protein CBL-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

DNRLSRHIHHVESVPSRDPPMPLEAWCPRDVFGTNQLVGCRLLGEGSPKPGITASSNVNGRHSRVGSDPVLMRKHRRHDLPLEGAKVFSNGHLGSEEYDVPPRLSPPPPVTTLLPSIKCTGPLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBLB(868)

General description

CBLB (Cbl proto-oncogene B) is an E3 ubiquitin protein ligase belonging to the CBL protein family. It is mapped to human chromosome 3q13.11. In mammals, there are three homologues of Cbl, namely c-Cbl, Cbl-b and Cbl-c. It is composed of highly conserved tyrosine kinase binding (TKB) domain and new gene (RING) finger catalytic domains.

Immunogen

E3 ubiquitin-protein ligase CBL-B recombinant protein epitope signature tag (PrEST)

Application

Anti-CBLB antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CBLB (Cbl proto-oncogene B) plays a crucial role in the regulation of immune cell activation. Both the domains perform different activities in ubiquitin ligase activity, such as TKB domain interacts directly with phosphorylated tyrosine-containing proteins through Src homology (SH) 2 domains and RING finger domain binds to E2 ubiquitin-conjugating. CBLB is phosphorylated upon T cell receptor (TCR) stimulation and is involved in TCR-mediated intracellular signaling pathways enzymes. Similarly, phosphorylated CBLB is recruited to epidermal growth factor receptor (EGFR) upon EGF stimulation and inhibits EGF-induced cell growth. CBLB participates in controlling T-cell activation thresholds by negative regulation of Vav, a GDP/GTP exchange factor. The molecular mechanism involves CBLB-mediated degradation of p85 regulatory subunit of phosphatidylinositol 3-kinase, an upstream regulator of Vav. CBLB is associated with an autoimmune disease of the central nervous system, multiple sclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73955

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Ettenberg et al.
Oncogene, 18(10), 1855-1866 (1999-03-23)
The role of cbl-b in signaling by the epidermal growth factor receptor (EGFR) was studied and compared with c-cbl. We demonstrate in vivo, that cbl-b, like c-cbl, is phosphorylated and recruited to the EGFR upon EGF stimulation and both cbl
D Fang et al.
The Journal of biological chemistry, 276(7), 4872-4878 (2000-11-23)
Cbl-b is implicated in setting the threshold of T lymphocyte activation. In Cbl-b-deficient T cells, the activation of Vav, a guanine nucleotide exchange factor, is significantly enhanced. The molecular mechanism underlying Cbl-b-regulated Vav activation was unclear. Here it is shown
Jezabel Varadé et al.
Multiple sclerosis (Houndmills, Basingstoke, England), 18(7), 959-965 (2011-12-24)
Ten genes previously showing different evidence of association with multiple sclerosis have been selected to validate. Eleven polymorphisms were genotyped with the iPLEX™ Sequenom in a well-powered collection of Spanish origin including 2863 multiple sclerosis cases and 2930 controls. Replication
C Elly et al.
Oncogene, 18(5), 1147-1156 (1999-02-18)
Cbl-b, a mammalian homolog of Cbl, consists of an N-terminal region (Cbl-b-N) highly homologous to oncogenic v-Cbl, a Ring finger, and a C-terminal region containing multiple proline-rich stretches and potential tyrosine phosphorylation sites. In the present study, we demonstrate that
Tomoki Abe et al.
Endocrine journal, 61(6), 529-538 (2014-03-13)
Obesity causes type 2 diabetes, atherosclerosis and cardiovascular diseases by inducing systemic insulin resistance. It is now recognized that obesity is related to chronic low-grade inflammation in adipose tissue. Specifically, activated immune cells infiltrate adipose tissue and cause inflammation. There

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service