Skip to Content
Merck
All Photos(2)

Documents

HPA019113

Sigma-Aldrich

Anti-SYNE1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-8B, Anti-ARCA1, Anti-C6orf98, Anti-CPG2, Anti-KIAA0796, Anti-MYNE1, Anti-Nesp1, Anti-Nesprin-1, Anti-SCAR8, Anti-SYNE-1B, Anti-dJ45H2.2, Anti-enaptin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SRDLESAMSRALPSEDEEGQDDKDFYLRGAVGLSGDHSALESQIRQLGKALDDSRFQIQQTENIIRSKTPTGPELDTSYKGYMKLLGECSSSIDSVKRLEHKLKEEEESLPGFVNLHSTETQTAGVIDRWEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYNE1(23345)

General description

The gene SYNE1 encoding synaptic nuclear envelope protein 1 is mapped to human chromosome 6q25. It belongs to family of nuclear membrane associated proteins, referred as nesprins. SYNE1 is ubiquitously expressed. The protein localizes at the nuclear membrane and contains amino-terminal α-actinin type actin-binding domain, a coiled coil rod and a carboxyl-terminal transmembrane domain. SYNE1 is commonly called as Nesprin-1.

Immunogen

Nesprin-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SYNE1 (synaptic nuclear envelope protein 1) is responsible for linking nucleus to the actin cytoskeleton. It interacts with the nuclear envelope proteins, emerin and lamin A. Single nucleotide polymorphism in SYNE1 is associated with menstrual migraine. Mutations in SYNE1 are linked with autosomal recessive cerebellar ataxia. It is also associated with the pathogenesis of Emery Dreifuss muscular dystrophy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74697

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid J Rodriguez-Acevedo et al.
The journal of headache and pain, 15, 62-62 (2014-10-16)
Menstrual migraine (MM) encompasses pure menstrual migraine (PMM) and menstrually-related migraine (MRM). This study was aimed at investigating genetic variants that are potentially related to MM, specifically undertaking genotyping and mRNA expression analysis of the ESR1, PGR, SYNE1 and TNF
François Gros-Louis et al.
Nature genetics, 39(1), 80-85 (2006-12-13)
The past decade has seen great advances in unraveling the biological basis of hereditary ataxias. Molecular studies of spinocerebellar ataxias (SCA) have extended our understanding of dominant ataxias. Causative genes have been identified for a few autosomal recessive ataxias: Friedreich's
John M K Mislow et al.
FEBS letters, 525(1-3), 135-140 (2002-08-07)
Nesprin-1alpha is a spectrin repeat (SR)-containing, transmembrane protein of the inner nuclear membrane, and is highly expressed in muscle cells. A yeast two-hybrid screen for nesprin-1alpha-interacting proteins showed that nesprin-1alpha interacted with itself. Blot overlay experiments revealed that nesprin-1alpha's third
Surayya Taranum et al.
International journal of cell biology, 2012, 736524-736524 (2012-04-21)
Nesprin-1 is a giant tail-anchored nuclear envelope protein composed of an N-terminal F-actin binding domain, a long linker region formed by multiple spectrin repeats and a C-terminal transmembrane domain. Based on this structure, it connects the nucleus to the actin
V C Padmakumar et al.
Experimental cell research, 295(2), 330-339 (2004-04-20)
Enaptin belongs to a family of recently identified giant proteins that associate with the F-actin cytoskeleton as well as the nuclear membrane. It is composed of an N-terminal alpha-actinin type actin-binding domain (ABD) followed by a long coiled coil rod

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service