Skip to Content
Merck
All Photos(8)

Key Documents

HPA019098

Sigma-Aldrich

Anti-AIMP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-JTV1 antibody produced in rabbit, Anti-Multisynthetase complex auxiliary component p38, Anti-Protein JTV-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

APLRVELPTCMYRLPNVHGRSYGPAPGAGHVQEESNLSLQALESRQDDILKRLYELKAAVDGLSKMIQTPDADLDVTNIIQADEPTTLTTNAL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JTV1(7965)

General description

The gene AIMP2 (aminoacyl tRNA synthase complex-interacting multifunctional protein 2) is mapped to human chromosome 7p22. AIMP2 is also referred as multisynthase complex auxiliary component p38.

Immunogen

Multisynthetase complex auxiliary component p38 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AIMP2 (aminoacyl tRNA synthase complex-interacting multifunctional protein 2) is crucial for assembly of the subunits involved in formation of tRNA synthetase complex. It associates with different tRNA synthetases (Arginine-RS, Glutamine-RS, Aspartate-RS, Lysine-RS and bifunctional GlutamateProline-RS). Presence of AIMP2 induces cell death. AIMP2 is up-regulated in brains of autosomal-recessive juvenile parkinsonism and idiopathic Parkinson′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74922

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Quevillon et al.
Journal of molecular biology, 285(1), 183-195 (1999-01-08)
In eukaryotes, from fly to human, nine aminoacyl-tRNA synthetases contribute a multienzyme complex of defined and conserved structural organization. This ubiquitous multiprotein assemblage comprises a unique bifunctional aminoacyl-tRNA synthetase, glutamyl-prolyl-tRNA synthetase, as well as the monospecific isoleucyl, leucyl, glutaminyl, methionyl
Haiping Dai et al.
PloS one, 10(11), e0139663-e0139663 (2015-11-26)
Primary mediastinal B-Cell lymphoma (PMBL) is a recently defined entity comprising ~2-10% non-Hodgkin lymphomas (NHL). Unlike most NHL subtypes, PMBL lacks recurrent gene rearrangements to serve as biomarkers or betray target genes. While druggable, late chemotherapeutic complications warrant the search
Han Seok Ko et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(35), 7968-7978 (2005-09-02)
Autosomal-recessive juvenile parkinsonism (AR-JP) is caused by loss-of-function mutations of the parkin gene. Parkin, a RING-type E3 ubiquitin ligase, is responsible for the ubiquitination and degradation of substrate proteins that are important in the survival of dopamine neurons in Parkinson's

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service