Skip to Content
Merck
All Photos(10)

Documents

HPA018871

Sigma-Aldrich

Anti-SERPINB1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EI, Anti-LEI, Anti-Leukocyte elastase inhibitor, Anti-M/NEI, Anti-Monocyte/neutrophil elastase inhibitor, Anti-Serpin B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

GNWKDKFMKEATTNAPFRLNKKDRKTVKMMYQKKKFAYGYIEDLKCRVLELPYQGEELSMVILLPDDIEDESTGLKKIEEQLTLEKLHEWTKPENLDFIEVNVSLPRFK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINB1(1992)

General description

The gene SERPINB1 (leukocyte elastase inhibitor) is mapped to human chromosome 6p25. It belongs to SERPIN superfamily and SERPINB family. SERPINB1 is present in all bone marrow leukocytes.

Immunogen

Leukocyte elastase inhibitor recombinant protein epitope signature tag (PrEST)

Application

Anti-SERPINB1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SERPIN superfamily proteins are mainly involved in suppressing proteolytic activities of serine proteases. SERPINB1 is an inhibitor of granzyme-H (GzmH). The granzyme pathway is important for cytotoxic lymphocytes to remove virus-infected and tumor cells. SERPINB1 suppresses GzmH-mediated cytotoxicity. Presence of SERPINB1 restricts production of NETs (neutrophil extracellular trap), thereby regulating NETosis, a programmed death pathway. It is up-regulated in patients with ulcerative colitis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74613

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Gallardo et al.
Molecular cancer, 12, 142-142 (2013-11-21)
JAK-STAT signaling through the JAK2V617F mutation is central to the pathogenesis of myeloproliferative neoplasms (MPN). However, other events could precede the JAK2 mutation. The aim of this study is to analyze the phenotypic divergence between polycytemia vera (PV) and essential
Christoph Willmes et al.
Oncotarget, 7(9), 10117-10132 (2016-01-23)
Despite of highly effective new therapeutic strategies, chemotherapy still is an important treatment option in metastatic melanoma. Since predictors of chemotherapy response are rare, drugs and regimens are currently chosen arbitrarily. The present study was aimed at the identification of
Li Wang et al.
Journal of immunology (Baltimore, Md. : 1950), 190(3), 1319-1330 (2012-12-28)
The granzyme/perforin pathway is a major mechanism for cytotoxic lymphocytes to eliminate virus-infected and tumor cells. The balance between activation and inhibition of the proteolytic cascade must be tightly controlled to avoid self damage. Granzyme H (GzmH) is constitutively expressed
Kazuhiko Uchiyama et al.
American journal of physiology. Gastrointestinal and liver physiology, 302(10), G1163-G1170 (2012-03-17)
Serpin B1 is a monocyte neutrophil elastase (NE) inhibitor and is one of the most efficient inhibitors of NE. In the present study, we investigated the role of serpin B1 in the pathogenesis of ulcerative colitis by using clinical samples
Kalamo Farley et al.
Journal of immunology (Baltimore, Md. : 1950), 189(9), 4574-4581 (2012-09-25)
NETosis (neutrophil extracellular trap [NET] generation), a programmed death pathway initiated in mature neutrophils by pathogens and inflammatory mediators, can be a protective process that sequesters microbes and prevents spread of infection, but it can also be a pathological process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service