Skip to Content
Merck
All Photos(4)

Documents

HPA018794

Sigma-Aldrich

Anti-PDLIM7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LIM mineralization protein, Anti-LMP, Anti-PDZ and LIM domain protein 7, Anti-Protein enigma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HLTGTEFMQDPDEEHLKKSSQVPRTEAPAPASSTPQEPWPGPTAPSPTSRPPWAVDPAFAERYAPDKTSTVLTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM7(9260)

General description

The gene PDLIM7 (PDZ and LIM domain protein-7) is mapped to human chromosome 5q35.3. It belongs to PDLIM family. PDLIM7 has PDZ (Discs-large homologous regions) domain and LIM (Lin11, Isl-1 & Mec-3) domains. PDLIM7 is expressed ubiquitously in human tissues, including leukocytes, spleen, lung, placenta, fetal liver, skeletal muscle, bone marrow and heart. PDLIM7 is also called as LMP1 (LIM mineralization protein).

Immunogen

PDZ and LIM domain protein 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Human PDZ and LIM domain protein-7 (PDLIM7) induces bone formation upon overexpression in fetal rat calvarial osteoblast cultures. PDLIM7 binds CBLC (E3 ubiquitin-protein ligase) and blocks CBLC-mediated degradation of receptor tyrosine kinase RETMEN2A, thereby enhancing ERK (Extracellular signal-regulated kinase) activation via RETMEN2A. Similarly, PDLIM7 binds with SMURF1 (Smad ubiquitin regulatory factor 1) and blocks ubiquitination of Smads (Mothers against decapentaplegic homolog), thus causing potentiation of bone morphogenetic protein activity. PDLIM7 is down-regulated in osteosarcoma tissues. Presence of PDLIM7 suppresses cell proliferation and invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74510

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dunja Niedrist et al.
European journal of human genetics : EJHG, 17(8), 1086-1091 (2009-02-19)
On the basis of the Human Cytogenetic Database, a computerized catalog of the clinical phenotypes associated with cytogenetically detectable human chromosome aberrations, we collected from the literature 102 cases with chromosomal aberrations and split hand/foot malformation or absent fingers/toes. Statistical
Huiwen Liu et al.
International journal of molecular sciences, 15(4), 7037-7048 (2014-04-26)
Osteosarcoma (OS), also known as osteogenic sarcoma, is the most common primary malignancy of bone tumor in children and adolescents. However, its underlying molecular pathogenesis is still only vaguely understood. Recently, LIM mineralization protein-1 (LMP-1) was reported to be an
Stephen C Kales et al.
PloS one, 9(1), e87116-e87116 (2014-01-28)
The Cbl proteins (Cbl, Cbl-b, and Cbl-c) are a highly conserved family of RING finger ubiquitin ligases (E3s) that function as negative regulators of tyrosine kinases in a wide variety of signal transduction pathways. In this study, we identify a
Sreedhara Sangadala et al.
Molecular and cellular biochemistry, 385(1-2), 145-157 (2013-10-01)
Development and repair of the skeletal system and other organs are highly dependent on precise regulation of the bone morphogenetic protein (BMP) pathway. The use of BMPs clinically to induce bone formation has been limited in part by the requirement
Yunshan Liu et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 17(3), 406-414 (2002-03-05)
Rat LIM mineralization protein 1 (LMP-1, an LIM domain protein) mediates bone morphogenetic protein 6 (BMP-6) induction of bone nodule formation in fetal rat calvarial osteoblast (ROB) cultures. We have isolated the complementary DNA (cDNA) for the human homologue of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service