Skip to Content
Merck
All Photos(4)

Key Documents

HPA018499

Sigma-Aldrich

Anti-USP4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deubiquitinating enzyme 4, Anti-Ubiquitin carboxyl-terminal hydrolase 4, Anti-Ubiquitin thioesterase 4, Anti-Ubiquitin-specific-processing protease 4, Anti-Ubiquitous nuclear protein homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NMVVADVYNHRFHKIFQMDEGLNHIMPRDDIFVYEVCSTSVDGSECVTLPVYFRERKSRPSSTSSASALYGQPLLLSVPKHKLTLESLYQAVCDRISRYVKQPLPDEFGSSPL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP4(7375)

General description

The gene USP4 (ubiquitin carboxyl-terminal hydrolase-4) is mapped to human chromosome 3p21.3. The protein is present in the cytoplasm and the nucleus.

Immunogen

Ubiquitin carboxyl-terminal hydrolase 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

USP proteins are mainly responsible for deubiquitination of target proteins. Ubiquitin carboxyl-terminal hydrolase-4 (USP4) binds and deubiquitinates retinoic acid-inducible gene 1 (RIG1), thereby positively regulating RIG1-mediated antiviral response. USP4 deubiquitinates TRAF6 (TNF receptor-associated factor 6) and prevents the activation of NF (Nuclear factor)-κB and AP-1 (Activator protein-1) transcription factors, and subsequent innate immune activation. USP4 mRNA is elevated in small cell tumors and adenocarcinomas of the lung. USP4 is up-regulated in head and neck squamous cell carcinoma (HNSCC) and promotes tumor necrosis factor-α-induced apoptosis in HNSCC.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86202

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruirong Lin et al.
FEBS letters, 591(12), 1677-1686 (2017-05-10)
CD4+ CD25+ regulatory T (Treg) cells comprise a unique subset of T cells required for maintaining immune homeostasis. However, the molecular mechanisms associated with the functional variety of Treg cells are not fully delineated. In the present study, we demonstrate
Eun Joo Song et al.
Genes & development, 24(13), 1434-1447 (2010-07-03)
The spliceosome, a dynamic assembly of proteins and RNAs, catalyzes the excision of intron sequences from nascent mRNAs. Recent work has suggested that the activity and composition of the spliceosome are regulated by ubiquitination, but the underlying mechanisms have not
D A Gray et al.
Oncogene, 10(11), 2179-2183 (1995-06-01)
The murine Unp proto-oncogene encodes a nuclear ubiquitin protease whose overexpression leads to oncogenic transformation of NIH3T3 cells. We have cloned cDNAs originating from the human homolog of this gene, designated herein as Unph (Unp, human), and have used these
Xiaozhi Hou et al.
FEBS letters, 587(4), 311-316 (2013-01-15)
Head and neck squamous cell carcinoma (HNSCC) is a common type of cancer. Better understanding of molecular aberrations associated with HNSCC might identify new diagnostic and therapeutic strategies for this disease. In this study, we found ubiquitin-specific protease 4 (USP4)
Lijuan Wang et al.
Journal of virology, 87(8), 4507-4515 (2013-02-08)
Protein ubiquitination plays an essential role in the regulation of retinoic acid-inducible gene I (RIG-I) activation and the antiviral immune response. However, the function of the opposite process of deubiquitination in RIG-I activation remains elusive. In this study, we have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service