Skip to Content
Merck
All Photos(1)

Documents

HPA018325

Sigma-Aldrich

Anti-UPF3A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Nonsense mRNA reducing factor 3A, Anti-Regulator of nonsense transcripts 3A, Anti-Up-frameshift suppressor 3 homolog A, Anti-hUpf3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KARPMEGSLEEPQETSHSGSDKEHRDVERSQEQESEAQRYHVDDGRRHRAHHEPERLSRRSEDEQRWGKGPGQDRGKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UPF3A(65110)

General description

Up-frameshift suppressor 3 homolog A (UPF3A) shuttles between nucleus and cytoplasm. In humans there are two paralogs of UPF3, UPF3A and UPF3B. UPF3A transcript is highly expressed in testis, uterus, muscle, fetal brain and spinal cord.

Immunogen

Regulator of nonsense transcripts 3A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Up-frameshift suppressor-3 homolog A (UPF3A) is involved in nonsense-mediated decay (NMD), an RNA decay pathway. A mutant of UPF3A which is unable to exit the nucleus does not participate in nonsense-mediated RNA decay pathway. UPF3A levels are tightly regulated in cells by its destabilization via UPF3B-dependent mechanism. UPF3A forms a complex with RNA-binding protein 8A (Y14), protein mago nashi homolog (MAGOH), eukaryotic initiation factor 4A-III (eIF4AIII) and barentsz (BTZ). Together the complex is required for NMD pathway. UPF3A interacts specifically with spliced mRNAs. It is also a component of exon-exon junction complex and is important for translation efficiency.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73979

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wai-Kin Chan et al.
Nature structural & molecular biology, 16(7), 747-753 (2009-06-09)
Nonsense-mediated decay (NMD) is an RNA decay pathway that downregulates aberrant mRNAs and a subset of normal mRNAs. The regulation of NMD is poorly understood. Here we identify a regulatory mechanism acting on two related UPF (up-frameshift) factors crucial for
V N Kim et al.
Science (New York, N.Y.), 293(5536), 1832-1836 (2001-09-08)
Nonsense-mediated messenger RNA (mRNA) decay, or NMD, is a critical process of selective degradation of mRNAs that contain premature stop codons. NMD depends on both pre-mRNA splicing and translation, and it requires recognition of the position of stop codons relative
G Serin et al.
Molecular and cellular biology, 21(1), 209-223 (2000-12-13)
Nonsense-mediated mRNA decay (NMD), also called mRNA surveillance, is an important pathway used by all organisms that have been tested to degrade mRNAs that prematurely terminate translation and, as a consequence, eliminate the production of aberrant proteins that could be
Guramrit Singh et al.
Molecular cell, 27(5), 780-792 (2007-09-07)
The nonsense-mediated mRNA decay (NMD) pathway rids eukaryotic cells of mRNAs with premature termination codons. There is contradictory evidence as to whether mammalian NMD is a nuclear or a cytoplasmic process. Here, we show evidence that NMD in human cells
Malwina Michalak et al.
International journal of molecular sciences, 21(15) (2020-07-29)
DNA mismatch repair-deficient colorectal cancers (CRCs) accumulate numerous frameshift mutations at repetitive sequences recognized as microsatellite instability (MSI). When coding mononucleotide repeats (cMNRs) are affected, tumors accumulate frameshift mutations and premature termination codons (PTC) potentially leading to truncated proteins. Nonsense-mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service