Skip to Content
Merck
All Photos(4)

Key Documents

HPA018254

Sigma-Aldrich

Anti-SPATA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Spermatogenesis-associated protein 3, Anti-Testis and spermatogenesis cell-related protein 1, Anti-Testis spermatocyte apoptosis- related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MKKVKKKRSEARRHRDSTSQHASSNSTSQQPSPESTPQQPSPESTPQHSSLETTSRQPAFQALPAPEIRRSSCCLLSPDAN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPATA3(130560)

General description

The gene SPATA3 (spermatogenesis-associated protein 3) has been mapped to human chromosome 2q37. The gene encodes a testis-specific protein. In rat model, SPATA3 is identified as a new member of HSP40 (Heat shock protein 40) protein family since the sequence contains the highly conserved J domain. The protein is localized in the cytoplasm.

Immunogen

Spermatogenesis-associated protein 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-SPATA3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Spermatogenesis-associated protein 3 (SPATA3) may play role in spermatogenesis cell apoptosis. In mouse model, SPATA3 is down-regulated in male germ cells post X-ray irradiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74241

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and Localization of pEGFP-C3/Tsarg1 Fusion Protein in GC-1 spg Cells.
Yi Z, et al.
Practical preventive medicine (Shiyong Yufang Yixue), 8 (2010)
Adel Driss et al.
Malaria journal, 10, 271-271 (2011-09-21)
The influence of host genetics on susceptibility to Plasmodium falciparum malaria has been extensively studied over the past twenty years. It is now clear that malaria parasites have imposed strong selective forces on the human genome in endemic regions. Different
Hong-Mei Yang et al.
DNA sequence : the journal of DNA sequencing and mapping, 16(3), 166-172 (2005-09-09)
Beginning with a mouse gene mTSARG3, which was related to apoptosis of spermatogenic cells, bioinformatics was applied and a predicted novel rat gene full-length cDNA sequence was attained. Gene-specific primers were designed for PCR in rat testis cDNA library. A
Fozia J Shah et al.
Reproductive biology and endocrinology : RB&E, 7, 130-130 (2009-11-21)
Irradiation or chemotherapy that suspend normal spermatogenesis is commonly used to treat various cancers. Fortunately, spermatogenesis in many cases can be restored after such treatments but knowledge is limited about the re-initiation process. Earlier studies have described the cellular changes
Linn Fagerberg et al.
Molecular & cellular proteomics : MCP, 13(2), 397-406 (2013-12-07)
Global classification of the human proteins with regards to spatial expression patterns across organs and tissues is important for studies of human biology and disease. Here, we used a quantitative transcriptomics analysis (RNA-Seq) to classify the tissue-specific expression of genes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service