Skip to Content
Merck
All Photos(3)

Key Documents

HPA018207

Sigma-Aldrich

Anti-ILDR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Immunoglobulin-like domain-containing receptor 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LEYLEKELRNLNLAQPLPPDLKGRFGHPCSMLSSLGSEVVERRIIHLPPLIRDLSSSRRTSDSLHQQWLTPIPSRPWDLREGRSHHHYPDFHQELQDRGPKSWALERRELDPSWSGRHRSSRLNGSPIHWSDRDSLSDVPS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ILDR1(286676)

General description

The gene immunoglobulin-like domain-containing receptor-1 (ILDR1) is mapped to human chromosome 3q13.33. ILDR1 is expressed in prostate, testis, pancreas, kidney, liver, and heart. The gene encodes three variants, two with membrane localization and one as cytoplasmic. The cytoplasmic variant is only detected in lymphoma samples.

Immunogen

Immunoglobulin-like domain-containing receptor 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Loss of function mutations in immunoglobulin-like domain-containing receptor-1 (ILDR1) leads to autosomal-recessive hearing impairment in humans. ILDR1 is present at the tricellular contacts of epithelial cells in mouse model. The protein is important for recruitment of tricellulin to tricellular contacts which helps in the establishment of a strong barrier of epithelium. However, ILDR1 is not significant for tricellulin recruitment in the auditory and vestibular sensory epithelia or in the stria vascularis, but is required for tricellilin retention and integrity of tricellular tight junctions which is essential for normal hearing.[1] ILDR1 is expressed in cholecystokinin cells. It regulates the release of cholecystokinin gastrointestinal hormone in the presence of lipoproteins and intestinally absorbed fatty acids. ILDR1 is up-regulated in myelodysplastic syndrome, refractory anemia with excess blasts.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72904

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khushnooda Ramzan et al.
European journal of medical genetics, 57(6), 253-258 (2014-04-29)
Hearing impairment is the common human sensorineural disorder and is a genetically heterogeneous phenotype for which more than 100 genomic loci have been mapped so far. ILDR1 located on chromosome 3q13.33, encodes a putative transmembrane receptor containing an immunoglobulin-like domain.
Eva L Morozko et al.
Human molecular genetics, 24(3), 609-624 (2014-09-14)
In the mammalian inner ear, bicellular and tricellular tight junctions (tTJs) seal the paracellular space between epithelial cells. Tricellulin and immunoglobulin-like (Ig-like) domain containing receptor 1 (ILDR1, also referred to as angulin-2) localize to tTJs of the sensory and non-sensory
Rashmi Chandra et al.
The Journal of clinical investigation, 123(8), 3343-3352 (2013-07-19)
Cholecystokinin (CCK) is a satiety hormone produced by discrete enteroendocrine cells scattered among absorptive cells of the small intestine. CCK is released into blood following a meal; however, the mechanisms inducing hormone secretion are largely unknown. Ingested fat is the
Helena Hauge et al.
Biochemical and biophysical research communications, 323(3), 970-978 (2004-09-24)
A not yet described gene was identified in a lymphoma specific subtraction strategy. Three splice variants of this gene denoted immunoglobulin-like domain containing receptor 1 (ILDR1) were identified and characterized. Cellular localization studies showed membrane-association for two of the variants
Tomohito Higashi et al.
Journal of cell science, 126(Pt 4), 966-977 (2012-12-15)
Tricellular tight junctions (tTJs) seal the extracellular space at tricellular contacts (TCs), where the corners of three epithelial cells meet. To date, the transmembrane proteins tricellulin and lipolysis-stimulated lipoprotein receptor (LSR) are known to be molecular components of tTJs. LSR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service