Skip to Content
Merck
All Photos(2)

Key Documents

HPA018178

Sigma-Aldrich

Anti-SLC30A4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Solute carrier family 30 member 4, Anti-Zinc transporter 4, Anti-ZnT-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PSHLNVDYIKEALMKIEDVYSVEDLNIWSLTSGKSTAIVHIQLIPGSSSKWEEVQSKANHLLLNTFGMYRCTIQLQSYRQEVDRTCANCQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC30A4(7782)

General description

The gene zinc transporter-4 (ZNT4) is mapped to human chromosome 15q21.1. ZNT4 belongs to family of zinc transporters (ZNTs). ZNTs have six transmembrane domains and histidine rich loops which connect these domains. ZNT4 is ubiquitously expressed and is localized in the plasma membrane and lysosomes.

Immunogen

Zinc transporter 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Zinc transporter-4 (ZNT4) transports Zn from the cytosol to lysosomes and endosomes. siRNA-mediated down-regulation of ZNT4 eliminates the Zn-induced lysosomal swelling and Zn retention in TRPML1 (mucolipin-1)-deficient HeLa cells. Premature termination codon in ZNT4 decreases the transport of Zn into milk in mouse model. ZNT4 is involved in the pathological process which causes β-amyloid peptide containing senile plaque formation in Alzheimer′s disease. ZNT4 is down-regulated in prostate benign prostatic hyperplasis and tumor samples.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73259

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard D Palmiter et al.
Pflugers Archiv : European journal of physiology, 447(5), 744-751 (2003-05-16)
All of the members of this family are thought to facilitate zinc efflux from the cytoplasm either into various intracellular compartments (endosomes, secretory granules, synaptic vesicles, Golgi apparatus, or trans-Golgi network) or across the plasma membrane. Thus, these transporters are
Silke Overbeck et al.
Journal of leukocyte biology, 83(2), 368-380 (2007-11-01)
Intracellular zinc homeostasis is strictly regulated by zinc binding proteins and zinc transporters. In the present study, we quantified in a first global view the expression of all characterized human zinc exporters (hZnT-1-9) in different leukocyte subsets in response to
Frances W J Beck et al.
The Prostate, 58(4), 374-381 (2004-02-18)
Altered zinc levels in prostate benign prostatic hyperplasia (BPH) and carcinoma is well documented. It is not known whether loss of zinc, necessary to restrain aggressive growth, results from loss of a single specific or multiple zinc transporters. Human prostate
Li-Hong Zhang et al.
Brain research bulletin, 77(1), 55-60 (2008-07-22)
The pathological key features of Alzheimer's disease (AD) are beta-amyloid peptide (Abeta)-containing senile plaques (SP) and neurofibrillary tangles. Previous studies have suggested that an extracellular elevation of the zinc concentration can initiate the deposition of Abeta and lead to the
Susan M Henshall et al.
Oncogene, 22(38), 6005-6012 (2003-09-05)
We have utilized oligonucleotide microarrays to identify novel genes of potential clinical and biological importance in prostate cancer. RNA from 74 prostate cancers and 164 normal body samples representing 40 different tissues were analysed using a customized Affymetrix GeneChip oligonucleotide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service