Skip to Content
Merck
All Photos(3)

Key Documents

HPA018173

Sigma-Aldrich

Anti-GRPEL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GrpE protein homolog 2, mitochondrial precursor, Anti-Mt-GrpE#2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EPEDQKLTLEKVFRGLLLLEAKLKSVFAKHGLEKLTPIGDKYDPHEHELICHVPAGVGVQPGTVALVRQDGYKLHGRTIRLARVEVAVES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRPEL2(134266)

General description

Heat shock protein-70 functions along with cochaperons, specifically J-domain proteins and nucleotide exchange factors (NEFs), which accelerate ATP hydrolysis and ADP-ATP exchange, respectively. The human cells express two mitochondrial NEF isoforms: Mitochondrial-GrpE protein homolog-1 (GRPEL1) and GRPEL2.

Immunogen

GrpE protein homolog 2, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mitochondrial-GrpE protein homolog-2 (GRPEL2) is up-regulated in active fibrovascular membranes. Using analytical ultracentrifugation and glutaraldehyde cross-linking GRPEL was shown to form dimer, which upon increasing protein concentration formed oligomers. The dimer can associate with DnaK and mitochondrial heat shock protein-70 (mtHSP70). mtHSP70 participates in protein folding, with help of GRPEL and mitochondrial DnaJ.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72942

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Schroda et al.
The Plant cell, 13(12), 2823-2839 (2001-12-26)
In eubacteria and mitochondria, Hsp70 chaperone activity is controlled by the nucleotide exchange factor GrpE. We have identified the chloroplastic GrpE homolog of Chlamydomonas, CGE1, as an approximately 26-kD protein coimmunoprecipitating with the stromal HSP70B protein. When expressed in Escherichia
Shigeo Yoshida et al.
The British journal of ophthalmology, 94(6), 795-801 (2009-11-19)
The purpose of this study was to generate a profile of genes expressed in preretinal fibrovascular membranes (FVMs) from patients with proliferative diabetic retinopathy. A PCR-amplified complementary DNA (cDNA) library was constructed using the RNAs isolated from FVMs obtained during
Nicholas J Hoogenraad et al.
Biochimica et biophysica acta, 1592(1), 97-105 (2002-08-23)
Most of our knowledge regarding the process of protein import into mitochondria has come from research employing fungal systems. This review outlines recent advances in our understanding of this process in mammalian cells. In particular, we focus on the characterisation
Cristiano L P Oliveira et al.
Archives of biochemistry and biophysics, 449(1-2), 77-86 (2006-04-04)
GrpE acts as a nucleotide exchange factor for the Hsp70 chaperone system. Only one GrpE isoform is present in Escherichia coli, but for reasons not yet well understood, two GrpE isoforms have been found in mammalian mitochondria.Therefore, studies aimed at
W Voos et al.
Molecular and cellular biology, 14(10), 6627-6634 (1994-10-01)
We characterized a 24-kDa protein associated with matrix hsp70 (mt-hsp70) of Neurospora crassa and Saccharomyces cerevisiae mitochondria. By using specific antibodies, the protein was identified as MGE, a mitochondrial homolog of the prokaryotic heat shock protein GrpE. MGE extracted from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service