Skip to Content
Merck
All Photos(4)

Key Documents

HPA018129

Sigma-Aldrich

Anti-ZFAND5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AN1-type zinc finger protein 5, Anti-Zinc finger A20 domain-containing protein 2, Anti-Zinc finger protein 216

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
7
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TASGSNSPTSDSASVQRADTSLNNCEGAAGSTSEKSRNVPVAALPVTQQMTEMSISREDKITTPKTEVSEPVVTQPSPSVSQPSTSQSEEKAPELPKPKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFAND5(7763)

General description

The gene has been mapped to human chromosome 9q13-q21. AN1-type zinc finger protein (ZFAND5) contains an A20-like zinc finger domain and AN1-like domain at the N- and C-terminus respectively. The two domains interact with each other. The protein is predominantly expressed in brain and skeletal muscles. Protein localizes largely in the cytoplasm and to a lesser extent in the nucleus of COS-7 cells.

Immunogen

AN1-type zinc finger protein 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AN1-type zinc finger protein (ZFAND5) regulates degradation of muscle proteins by binding to the polyubiquitin chains and associating with the 26S proteasome. ZFAND5-deficient mice exhibit resistance to muscle atrophy accompanied by abnormal accumulation of polyubiquitinylated proteins in the skeletal muscle. The protein interacts with inhibitor of nuclear factor-κ-B kinase subunit γ (IKKγ), receptor-interacting serine/threonine-protein kinase-1 (RIP) and TNF receptor-associated factor-6 (TRAF6). Overexpressed ZFAND5 inhibit RIP- and TRAF6-mediated NF-κ-B activation. Additionally, it could inhibit tumor necrosis factor (TNF), interleukin-1 (IL-1), and toll-like receptor 4 (TLR4) triggered NF-κ-B activation. Overexpression of the protein sensitizes cells to TNF-induced apoptosis. ZFAND5 is also an inhibitory factor for osteoclast differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73691

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and mutation analysis of a cochlear-expressed, zinc finger protein gene at the DFNB7/11 and dn hearing-loss loci on human chromosome 9q and mouse chromosome 19.
Scott DA, et al.
Gene, 215, 461-469 (1998)
G Sabatino et al.
Journal of biological regulators and homeostatic agents, 27(3), 729-738 (2013-10-25)
We tried to identify molecular markers in peripheral blood to predict high risk of aneurysm rupture. Extraction of the total population of peripheral blood mononuclear cell (PBMC) from total blood volume, total RNA extraction from PBMC and Agilent One Color
Amde Selassie Shifera
Biochemical and biophysical research communications, 396(3), 585-589 (2010-05-12)
Inhibitor of kappaB kinase (IKK) gamma (IKKgamma), also referred to as nuclear factor kappaB (NF-kappaB) essential modulator (NEMO), is an important component of the IKK complex. Following the exposure of cells to NF-kappaB-inducing stimuli, the IKK complex catalyzes the phosphorylation
Sarah Ressel et al.
Nucleic acids research, 52(9), 4872-4888 (2024-02-27)
microRNAs (miRNAs) regulate nearly all physiological processes but our understanding of exactly how they function remains incomplete, particularly in the context of viral infections. Here, we adapt a biochemical method (CLEAR-CLIP) and analysis pipeline to identify targets of miRNAs in
Akinori Hishiya et al.
The EMBO journal, 25(3), 554-564 (2006-01-21)
The ubiquitin-proteasome system (UPS) is critical for specific degradation of cellular proteins and plays a pivotal role on protein breakdown in muscle atrophy. Here, we show that ZNF216 directly binds polyubiquitin chains through its N-terminal A20-type zinc-finger domain and associates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service