Skip to Content
Merck
All Photos(4)

Key Documents

HPA017967

Sigma-Aldrich

Anti-HLA-DPA1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AL805913.4, Anti-DP(W3), Anti-DP(W4), Anti-HLA class II histocompatibility antigen, DP alpha chain precursor, Anti-HLA-SB alpha chain, Anti-MHC class II DP3-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RPTGEFMFEFDEDEMFYVDLDKKETVWHLEEFGQAFSFEAQGGLANIAILNNNLNTLIQRSNHTQATNDPPEVTVFPKEPVELGQPNTLICHIDKFFPPVLNVTWLCNGELVTEGVAESLFLPRTDYSFHKFHYLTF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HLA-DPA1(3113)

General description

Major histocompatibility complex, class II, DP α-1 (HLA-DPA1) belongs to the class II human leukocyte antigen (HLA) family and is expressed in Kupffer cells of the liver. The gene encoding it is localized on human chromosome 6.

Immunogen

HLA class II histocompatibility antigen, DP alpha chain precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-HLA-DPA1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Major histocompatibility complex, class II, DP α-1 (HLA-DPA1) is involved in viral antigen presentation to CD4+ T helper cells. Mutations in the gene encoding HLA-DPA1 has been associated with persistent hepatitis B virus infection.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72203

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Zhang et al.
Journal of virology, 87(22), 12176-12186 (2013-09-06)
Genetic polymorphisms of HLA-DP have been associated with hepatitis B virus (HBV) persistence. We aimed to determine the effect of HLA-DP polymorphisms on the generation of HBV mutations and their interactions on the outcomes of HBV infection. rs3077, rs3135021, rs9277535
Fabíola A Leite et al.
Pediatric blood & cancer, 61(11), 1940-1948 (2014-08-27)
Low expression of HLA class II antigens has been associated with more aggressive disease in several human malignancies including adult adrenocortical tumors (ACT), but their clinical relevance in pediatric ACT needs to be investigated. This study analyzed the expression profile
Martin E Baumdick et al.
Gastroenterology, 165(4), 946-962 (2023-07-17)
Ulcerative colitis (UC) is characterized by severe inflammation and destruction of the intestinal epithelium, and is associated with specific risk single nucleotide polymorphisms in HLA class II. Given the recently discovered interactions between subsets of HLA-DP molecules and the activating
Seisuke Kusano et al.
Journal of molecular biology, 426(17), 3016-3027 (2014-07-16)
The major allergen, Cry j 1, was isolated from Japanese cedar Cryptomeria japonica (Cry j) pollen and was shown to react with immunoglobulin E antibodies in the sera from pollinosis patients. We previously reported that the frequency of HLA-DP5 was
Jingtao Guo et al.
Cell research, 28(12), 1141-1157 (2018-10-14)
Human adult spermatogenesis balances spermatogonial stem cell (SSC) self-renewal and differentiation, alongside complex germ cell-niche interactions, to ensure long-term fertility and faithful genome propagation. Here, we performed single-cell RNA sequencing of ~6500 testicular cells from young adults. We found five

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service